Skip to Content
Merck
All Photos(6)

Key Documents

517127

Sigma-Aldrich

Dimethyl carbonate

greener alternative

anhydrous, ≥99%

Synonym(s):

DMC, Carbonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
Beilstein:
635821
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
90 °C (lit.)
vapor pressure:
18 mmHg ( 21.1 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.1 (vs air)

vapor pressure

18 mmHg ( 21.1 °C)

Assay

≥99%

form

liquid

expl. lim.

4.22-12.87 % (lit.)

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

<0.002% water
<0.005% water (100mL pkg)

color

APHA: <50

refractive index

n20/D 1.368 (lit.)

bp

90 °C (lit.)

mp

2-4 °C (lit.)

density

1.069 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=C(OC)OC

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3

InChI key

IEJIGPNLZYLLBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl carbonate (DMC) is a versatile, non-toxic, biodegradable reagent with tunable chemical reactivity. It serves as a green alternative to dimethyl sulfate or methyl halides and phosgene for methylation and carboxylation reactions.[1] DMC can be prepared by the base-catalyzed reaction between carbon dioxide and methanol.[2]

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Greener alternative to conventional solvents and chemicals. Click here for more information.

Application

Dimethyl carbonate has been used to dissolve poly (lactic-co-glycolic acid) (PLGA) to form PLGA-coated bioactive glass-ceramic scaffolds.[3]

Features and Benefits

Greener solvent

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterisation of plga-coated porous bioactive glass-ceramic scaffolds for subchondral bone tissue engineering.
O'Shea TM and Miao X.
Proceeding of the 9th International Symposium on Ceramic Materials and Components for Energy and Environmental Applications (2009)
Hao Sun et al.
Nature communications, 10(1), 3302-3302 (2019-07-26)
Rechargeable sodium metal batteries with high energy density could be important to a wide range of energy applications in modern society. The pursuit of higher energy density should ideally come with high safety, a goal difficult for electrolytes based on
Direct synthesis of dimethyl carbonate from carbon dioxide and methanol catalyzed by base.
Fang S and Fujimoto K.
Applied Catalysis A: General, 142(1), L1-L3 (1996)
Transesterification of cyclic carbonates to dimethyl carbonate using solid oxide catalyst at ambient conditions: environmentally benign synthesis.
Meenakshisundaram Sankar et al.
ChemSusChem, 3(5), 575-578 (2010-04-02)
Fusheng Liu et al.
Journal of hazardous materials, 174(1-3), 872-875 (2009-09-24)
An environmentally friendly strategy for methanolysis of polycarbonate (PC) to recover bisphenol A (BPA) and dimethyl carbonate (DMC) was developed in which PC could be methanolyzed in an ionic liquid without any acid or base catalyst under moderate conditions. The

Articles

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

1–3 of 3 Questions  
  1. What is the difference between anhydrous and battery grade? Can we use this in lithium and sodium ion batteries as a solvent for electrolyte?

    1 answer
    1. The difference between anhydrous and battery grade dimethyl carbonate (DMC) lies in their purity and specific testing. Battery grade DMC has an acid content test, while anhydrous grade does not. There is no indication that anhydrous grade can't be used as a solvent for electrolytes in lithium and sodium ion batteries; however, battery grade is more recommended if available.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service