Skip to Content
Merck
All Photos(2)

Documents

220892

Sigma-Aldrich

1,4-Butanediol diglycidyl ether

≥95%

Synonym(s):

1,4-Bis(2,3-epoxypropyloxy)butane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O4
CAS Number:
Molecular Weight:
202.25
Beilstein:
115238
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

~10 mmHg ( 20 °C)

Assay

≥95%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

266 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

C(CCOCC1CO1)COCC2CO2

InChI

1S/C10H18O4/c1(3-11-5-9-7-13-9)2-4-12-6-10-8-14-10/h9-10H,1-8H2

InChI key

SHKUUQIDMUMQQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Butanediol diglycidyl ether (BDDE) is a viscous liquid , which is hygroscopic in nature. BDDE is the most commonly used homobifunctional epoxide compound. BDDE may react with amines, hydroxyls, sulfhydryl groups to produce secondary amines, ether or thioether bonds respectively.

Application

BDDE is a cross linking agent used
  • for preparing amylose, xylan and hydroxyethyl-cellulose
  • to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells
  • to cross link hyaluronic acid into hydrogels
  • for the activation of soluble dextran polymers.
  • Amine or hydroxyl containing bis-epoxy supports are reacted with BDDE to form particles containing terminal epoxide group for immobilization reactions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Yao Chen et al.
Materials (Basel, Switzerland), 14(16) (2021-08-28)
(1) Background: Obesity is one of the most widespread chronic diseases and increases the risk of several other chronic diseases, especially type 2 diabetes. (2) Methods: Endobarrier is a new medical device what is worn in the small intestines for
Yasemin Udum et al.
Organic electronics, 15(5), 997-1001 (2014-05-13)
We have developed a hole-blocking layer for bulk-heterojunction solar cells based on cross-linked polyethylenimine (PEI). We tested five different ether-based cross-linkers and found that all of them give comparable solar cell efficiencies. The initial idea that a cross-linked layer is
Lennart Kenne et al.
Carbohydrate polymers, 91(1), 410-418 (2012-10-10)
Definitions and methods for the quantification of degree of modification and cross-linking in cross-linked hyaluronic acid (HA) hydrogels are outlined. A novel method is presented in which the HA hydrogel is degraded by the enzyme chondroitinase AC and the digest
The new face of fillers: why evidence and experience both count.
Hema Sundaram
Journal of drugs in dermatology : JDD, 10(9), 964-964 (2011-11-05)
David Stocks et al.
Journal of drugs in dermatology : JDD, 10(9), 974-980 (2011-11-05)
Hyaluronic acid (HA) gels are commonly injected into the skin to lift rhytides and to improve facial appearance. The different processes used in their manufacture and formulation yield products with unique physical characteristics that play an important role in predicting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service