Skip to Content
Merck
All Photos(3)

Key Documents

133507

Sigma-Aldrich

2-Amino-5-nitrothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3O2S
CAS Number:
Molecular Weight:
145.14
Beilstein:
126797
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

195-200 °C (dec.) (lit.)

solubility

95% ethanol: soluble 1g/150g at 20 °C
diethyl ether: soluble 1g/250g at 20 °C
chloroform: insoluble
water: slightly soluble

functional group

nitro

SMILES string

Nc1ncc(s1)[N+]([O-])=O

InChI

1S/C3H3N3O2S/c4-3-5-1-2(9-3)6(7)8/h1H,(H2,4,5)

InChI key

MIHADVKEHAFNPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-5-nitrothiazole(ANT) is a hypoxic radiosensitizing drug. ANT forms square-planar complex, trans-[PdCl2(ANT)2] with palladium in methanol.

Application

2-Amino-5-nitrothiazole was used as diazo component in the synthesis of monoazo disperse dyes. It was used as matrix during matrix-assisted laser desorption/ionization time-of-flight mass spectrometric study of oligonucleotide and protein.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Skov et al.
Radiation research, 112(2), 273-282 (1987-11-01)
Complexes of general formula [PtCl2(NH3)L] with one radiosensitizing ligand per platinum are compared with ligand L alone, complexes with two radiosensitizers per platinum [PtCl2L2], and their analogs with NH3 ligands, with respect to radiosensitizing properties and toxicity in CHO cells.
T Eric Ballard et al.
Bioorganic & medicinal chemistry letters, 20(12), 3537-3539 (2010-05-22)
Head group analogues of the antibacterial and antiparasitic drug nitazoxanide (NTZ) are presented. A library of 39 analogues was synthesized and assayed for their ability to suppress growth of Helicobacter pylori, Campylobacter jejuni, Clostridium difficile and inhibit NTZ target pyruvate:ferredoxin
2-amino-5-nitrothiazole.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 31, 71-77 (1983-07-01)
Evaluation of 2-amino-5-nitrothiazole as a hypoxic cell radiosensitizer.
S Rockwell et al.
Radiation research, 90(3), 575-585 (1982-06-01)
Synthesis and Spectral Properties of Hetaryl Monoazo dyes derived from 2-amino-5-nitrothiazole.
OTUTU JO, et al.
Orient. J. Chem., 27(4), 2011-2011 (1389)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service