All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8N2S
CAS Number:
Molecular Weight:
164.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
137-139 °C (lit.)
SMILES string
Cc1cccc2sc(N)nc12
InChI
1S/C8H8N2S/c1-5-3-2-4-6-7(5)10-8(9)11-6/h2-4H,1H3,(H2,9,10)
InChI key
GRIATXVEXOFBGO-UHFFFAOYSA-N
Related Categories
General description
2-Amino-4-methylbenzothiazole is the major degradation product of dufulin pesticide. Adsorption of 2-amino-4-methylbenzothiazole on colloidal silver particles has been investigated by surface enhanced Raman scattering technique.
Application
2-Amino-4-methylbenzothiazole was used in the synthesis of N-(4-methyl-1,3-benzothiazol-yl)-N-[(1,4,5,8-tetramethoxy-2-napthyl)methylidene]amine and Schiff′s base via condensation with 2-acetonaphthone.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jyotirmoy Sarkar et al.
The journal of physical chemistry. B, 109(47), 22536-22544 (2006-07-21)
The adsorption of 2-amino-4-methylbenzothiazole (2-AMBT) on colloidal silver particles has been investigated by a surface enhanced Raman scattering (SERS) study. The SERS spectra of the 2-AMBT molecule at varied adsorbate concentrations recorded in different time domains are compared with its
Hua Zi Wang et al.
Environmental science and pollution research international, 21(6), 4331-4342 (2013-12-11)
Dufulin is a newly developed antiviral agent (or pesticide) that activates systemic acquired resistance of plants. This pesticide is widely used in China to prevent abroad viral diseases in rice, tobacco and vegetables. In this study, the potential impacts such
Jikang Yoo et al.
Archives of pharmacal research, 31(2), 142-147 (2008-03-28)
A series of nine new compounds bridged by acyl groups at the 5,8-dihydroxyl group of DHNQ were synthesized and their cytotoxic activity against L1210 and P388 cancer cells was examined. Their antitumor action in mice bearing S-180 cells in the
SYNTHESIS, STRUCTURAL CHARACTERIZATION AND ANTIMICROBIAL ACTIVITIES OF SOME TRANSITION METAL COMPLEXES CONTAINING 2-ACETONAPHTHONE.
Mishra AP and Mishra DP.
International Journal of Pharmacy and Biological Sciences, 2(3), 430-439 (2011)
Fabio Gosetti et al.
Environmental science and pollution research international, 22(11), 8288-8295 (2014-12-23)
This paper reports the study of the photodegradation reactions that tricyclazole can naturally undergo, under the action of sunlight, in aqueous solutions of standard tricyclazole and of the commercial BEAM(TM) formulation. The analyses are carried out by ultra-high performance liquid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service