Skip to Content
Merck
All Photos(1)

Key Documents

658499

Sigma-Aldrich

2-Amino-4-(4-nitrophenyl)thiazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3O2S
CAS Number:
Molecular Weight:
221.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

283-287 °C

functional group

nitro

SMILES string

Nc1nc(cs1)-c2ccc(cc2)[N+]([O-])=O

InChI

1S/C9H7N3O2S/c10-9-11-8(5-15-9)6-1-3-7(4-2-6)12(13)14/h1-5H,(H2,10,11)

InChI key

RIKJWJIWXCUKQV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M B Mattammal et al.
Biomedical mass spectrometry, 9(9), 376-380 (1982-09-01)
Electron impact fragmentation of 2-methyl-4-(4-nitrophenyl)-thiazole and 2-amino-4-(4-nitrophenyl)-thiazoles were studied. Prominent fragment ions result from: (1) elimination of an NO2 radical and of a neutral NO molecule; (2) 1,2 cleavage of the thiazole ring in both compounds to give a phenoxythiirene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service