Merck
  • Quantification of Sorafenib in Human Serum by Competitive Enzyme-Linked Immunosorbent Assay.
Quantification of Sorafenib in Human Serum by Competitive Enzyme-Linked Immunosorbent Assay.
Biological & pharmaceutical bulletin (2015-11-03)
Tetsuya Saita, Yuta Yamamoto, Satoshi Noda, Makoto Shioya, Daiki Hira, Akira Andoh, Shin-Ya Morita, Tomohiro Terada, Masashi Shin
ABSTRACT
The multikinase inhibitor sorafenib has been used in the treatment of hepatocellular carcinoma, renal cell carcinoma, and differentiated thyroid carcinoma. Here we have demonstrated the production of the first specific antibody against sorafenib. Anti-sorafenib serum was obtained by immunizing mice with an antigen conjugated with bovine serum albumin and carboxylic modified 4-(4-aminophenoxy)-N-methyl-2-pyridinecarboxamide (AMPC) using the N-succinimidyl ester method. Enzyme labeling of sorafenib with horseradish peroxidase was similarly performed using carboxylic modified AMPC. A simple competitive enzyme-linked immunosorbent assay (ELISA) for sorafenib was developed using the principle of direct competition between sorafenib and the enzyme marker for anti-sorafenib antibody, which had been adsorbed by the plastic surface of a microtiter plate. Serum sorafenib concentrations lower than 0.04 µg/mL were reproducibly measurable using the ELISA. This ELISA was specific to sorafenib and showed very slight cross-reactivity (2.5%) with a major metabolite, sorafenib N-oxide. The values of serum sorafenib levels from 32 patients measured by this ELISA were comparable with those measured by HPLC, and there was a strong correlation between the values determined by the two methods (Y=1.016X-0.137, r=0.979). The specificity and sensitivity of the ELISA for sorafenib should provide a valuable new tool for use in therapeutic drug monitoring and pharmacokinetic studies of sorafenib.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Picolinamide, 98%