Skip to Content
Merck
All Photos(3)

Key Documents

S1679

Sigma-Aldrich

Sodium chloride

meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
solid
solubility:
water: soluble (358 g/l at 20 °C (68 °F))

grade

for analytical purposes

Quality Level

Agency

meets analytical specifications of BP
meets analytical specifications of Ph. Eur.
meets analytical specifications of USP

vapor pressure

1.33 (1.00 mmHg) at 865 °C hPa

Assay

99.0-100.5%

form

solid

quality

meets analytical specification of Ph. Eur., BP, USP

loss

≤0.5% loss on drying

pH

7

mp

801 °C (lit.)

solubility

water: soluble (358 g/l at 20 °C (68 °F))

application(s)

pharmaceutical (small molecule)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Na and K Intake from Lunches Served in a Japanese Company Cafeteria and the Estimated Improvement in the Dietary Na/K Ratio Using Low-Na/K Seasonings and Dairy to Prevent Hypertension.: This study investigates sodium and potassium intake from company cafeteria meals and explores the effectiveness of low-sodium and potassium-rich seasonings in improving dietary balance to reduce hypertension risk (Okuda et al., 2024).

Other Notes

Easily compare specifications for Sodium Chloride products with the Sodium Chloride specification table.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25
Koji Tanaka et al.
Nature communications, 6, 6337-6337 (2015-02-27)
Pore-forming toxins (PFT) are water-soluble proteins that possess the remarkable ability to self-assemble on the membrane of target cells, where they form pores causing cell damage. Here, we elucidate the mechanism of action of the haemolytic protein fragaceatoxin C (FraC)
Laurent Dortet et al.
Journal of clinical microbiology, 52(7), 2359-2364 (2014-04-25)
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in
Luc H Boudreau et al.
Blood, 124(14), 2173-2183 (2014-08-02)
Mitochondrial DNA (mtDNA) is a highly potent inflammatory trigger and is reportedly found outside the cells in blood in various pathologies. Platelets are abundant in blood where they promote hemostasis. Although lacking a nucleus, platelets contain functional mitochondria. On activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service