Merck
  • Expression of NAT2 in immune system cells and the relation of NAT2 gene polymorphisms in the anti-tuberculosis therapy in Mexican mestizo population.
Expression of NAT2 in immune system cells and the relation of NAT2 gene polymorphisms in the anti-tuberculosis therapy in Mexican mestizo population.
Molecular biology reports (2014-08-29)
R Salazar-González, R Gómez, S Romano-Moreno, S Medellín-Garibay, A Núñez-Ruíz, M Magaña-Aquino, R C Milán-Segovia, D P Portales-Pérez
ABSTRACT
Arylamine N-acetyltransferase 2 (NAT2) metabolizes isoniazid (INH) and Single Nucleotide Polymorphisms (SNP) responsible for its activity has been reported. The aim of this study in the Mexican mestizo population was to evaluate NAT2 expression at the protein level in immune cells, as well as the distribution and frequency of six NAT2 SNPs and their association with anti-TB therapy, by measuring the plasma levels of INH and Acetyl-INH (AcINH). We performed genotyping assays of NAT2 SNPs in 40 TB patients and 121 healthy volunteers by real-time PCR. A method for detecting NAT2 in immune cells using flow cytometry was developed. Plasma concentrations of INH and AcINH were obtained by HPLC in TB patients and the Metabolic Ratio (MR) was calculated. The phenotypes obtained in the healthy volunteers were as follows; 18.87 % of subjects had the rapid acetylator phenotype, 45.45 % had the intermediate phenotype and 39.66 % exhibited the slow acetylator phenotype. In the TB patient group, 35 % of patients had the rapid acetylator phenotype, 32.5 % were intermediate and 32.5 % showed the slow acetylator phenotype. A higher expression level of NAT2 in innate immune cells from TB patients compared to those from healthy volunteers was detected (P < 0.013). In TB patients the MR showed a bimodal distribution with an antimode of 0.7, which was used as a threshold value for acetylator classification. A high correspondence between the rapid and slow acetylator phenotype with MR was demonstrated. In conclusion, the 282C>T, 341T>C, 481C>T, 590G>A, 803A>G, 857G>A SNPs of NAT2 gene provides accurate for prediction of the acetylator phenotype in Mexican mestizo population. A statistical difference was found in frequency of rapid metabolizer phenotype, which was higher in TB patients. In addition, the expression of NAT2 protein in immune cells can lead to further studies related to its functional role in the innate immune response against M. tuberculosis and other xenobiotics metabolized by this enzyme.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium hexanesulfonate, ≥98% (elemental analysis)
Sigma-Aldrich
Sodium hexanesulfonate, BioXtra
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Sodium 1-hexanesulfonate solution, suitable for ion pair chromatography, concentrate, LiChropur, ampule
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)