Skip to Content
Merck
All Photos(4)

Documents

T8657

Sigma-Aldrich

Trichloroacetic acid

suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

vapor density

<1 (vs air)

vapor pressure

1 mmHg ( 51 °C)

Assay

≥99%

form

powder or flakes (or crystals)

technique(s)

electrophoresis: suitable

turbidity

≤3 NTU (1 g/ml)

refractive index

n20/D 1.62 (lit.)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

solubility

water: 1 g/mL, clear, colorless

density

1.62 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.002%

suitability

suitable for fixing solution (for IEF and PAGE gels)

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroacetic acid (TCA) is produced during the chlorination of water comprising natural organic material. It is a novel reagent used in clinical laboratories for deproteinization of body fluids. In addition, TCA is also used in the treatment of genital warts and acne scars.

Application

Trichloroacetic acid has been used:
  • for hydrolysis of DNA isolated from the cell
  • for precipitation of glutathione isolated from seminal plasma
  • as a fixative in microscopy

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teratogenic activity of trichloroacetic acid in the rat
Smith MK, et al.
Teratology, 40(5), 445-451 (1989)
Quantitation of DNA fragmentation with diphenylamine
Boraschi D and Maurizi G
Apoptosis-a laboratory manual of experimental methods, 153-161 (1998)
Determination of total protein in spinal fluid with sulphosalicylic acid and trichloroacetic acid
Meulemans O
Clinica Chimica Acta; International Journal of Clinical Chemistry, 5(5), 757-761 (1960)
Atmospheric pressure pulsed plasma induces cell death in photosynthetic organs via intracellularly generated ROS
Seol Y, et al.
Scientific reports, 7(1), 589-589 (2017)
Focal treatment of acne scars with trichloroacetic acid: chemical reconstruction of skin scars method
Lee J, et al.
Dermatologic Surgery : Official Publication For American Society For Dermatologic Surgery [Et Al.], 28(11), 1017-1021 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service