Skip to Content
Merck
All Photos(1)

Documents

E8008

Sigma-Aldrich

Ethylenediaminetetraacetic acid solution

0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

EDTA solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
Beilstein:
1716295
MDL number:
UNSPSC Code:
12352204
eCl@ss:
39030907
PubChem Substance ID:
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

0.02% in DPBS (0.5 mM)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

7.1-7.7

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid (EDTA) is an aminopolycarboxylic acid and a hexadentate ligand. It chelates with metal ions to form an octahedral complex especially with cations. Ethylenediaminetetraacetic acid (EDTA) is an anticoagulant, which prevents clotting of blood. It is implicated in platelet aggregation in pseudothrombocytopenia. EDTA chelates with calcium and is effective in solubilisation of the atheromatous plaques. Calcium disodium EDTA administration is effective in slowing down the progression of chronic kidney disease. It is a chelator used to remove calcium from cell washing and suspension media. EDTA reduces cell clumping.

Application

Ethylenediaminetetraacetic acid solution has been used to remove confluent cell monolayers in swine testicular (ST-cells) culture. It has also been used to detach embryonic stem cells (ESCs) from matrigel and primary human fibroblasts.

Physical form

Solution in Dulbecco′s phosphate buffered saline without calcium and magnesium

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exosomes from embryonic mesenchymal stem cells alleviate osteoarthritis through balancing synthesis and degradation of cartilage extracellular matrix
Wang Y, et al.
Stem Cell Research & Therapy, 8(1), 189-189 (2017)
The genotoxicity of physiological concentrations of chromium (Cr (III) and Cr (VI)) and cobalt (Co (II)): an in vitro study
Figgitt M, et al.
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 688(1-2), 53-61 (2010)
Ex vivo Lung bioengineering
Lung Stem Cells in the Epithelium and Vasculature, 151-151 (2015)
EDTA chelation therapy for cardiovascular disease: a systematic review
Seely DMR, et al.
BMC Cardiovascular Disorders, 5(1), 32-32 (2005)
Environmental fate and microbial degradation of aminopolycarboxylic acids
Bucheli-Witschel M and Egli T
FEMS Microbiology Reviews, 25(1), 69-106 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service