Skip to Content
Merck
All Photos(4)

Key Documents

T4885

Sigma-Aldrich

Trichloroacetic acid

≥99.0% (titration)

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

<1 (vs air)

Quality Level

vapor pressure

1 mmHg ( 51 °C)

Assay

≥99.0% (titration)

form

powder or crystals

refractive index

n20/D 1.62 (lit.)

pH

1 (at 25 °C, 81,7  g/L)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

solubility

H2O: 1 g/10 mL

density

1.62 g/mL at 25 °C (lit.)

functional group

carboxylic acid
chloro

storage temp.

2-8°C

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.
Trichloroacetic acid (TCA) can be used as a catalyst to synthesize:
  • β-enaminones via condensation reaction of 1,3-dicarbonyl compounds and amines.
  • 6-amino-3-methyl-1,4-diphenyl-1,4-dihydropyrano[2,3-c]pyrazole-5-carbonitrile derivatives via four-component condensation reaction of ethyl acetoacetate, phenylhydrazine, malononitrile, and aromatic aldehydes.
  • Tetrahydrobenzo[a]xanthen-11-ones by three-component reaction of aldehydes, 2-naphthol and dimedone.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expeditious, four-component synthesis of 1, 4-dihydropyrano [2, 3-c] pyrazole derivatives catalyzed by trichloroacetic acid or ceric sulfate
Jaberi ZK, et al.
Acta Chimica Slovenica, 60(1), 105-108 (2013)
Efficient, one-pot synthesis of tetrahydrobenzo [a] xanthen-11-ones and dibenzo [a, j] xanthenes using trichloroacetic acid as a solid heterogeneous catalyst under solvent-free conditions
Jaber ZK, et al.
E-Journal of Chemistry, 8(4), 1895-1899 (2011)
Efficient Synthesis of β-Enaminones and β-Enamino Esters Using Tris (Hydrogensulfato) Boron or Trichloroacetic Acid as Catalysts
Karimi-Jaberi Z and Takmilifard Z
European Chemical Bulletin, 2, 211-213 (2013)
Alan Mortensen et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 18(1), 55-62 (2012-10-17)
The ratio of the nitric oxide synthase (NOS) cofactor tetrahydrobiopterin (BH(4)) to its oxidized form dihydrobiopterin (BH(2)) has been suggested as an index of endothelial dysfunction. Consequently, much effort has been put into preserving the in vivo equilibrium between these
Inga Müller et al.
PloS one, 9(5), e97285-e97285 (2014-05-27)
Members of the MYC family are the most frequently deregulated oncogenes in human cancer and are often correlated with aggressive disease and/or poorly differentiated tumors. Since patients with MYCN-amplified neuroblastoma have a poor prognosis, targeting MYCN using small molecule inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service