Skip to Content
Merck
All Photos(1)

Key Documents

733466

Sigma-Aldrich

Acetonitrile

99.999% trace metals basis, electronic grade, battery grade

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Acetonitrile, electronic grade, 99.999% trace metals basis

grade

battery grade
electronic grade

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

99.999% trace metals basis

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

impurities

≤0.05% water
water

color

colorless

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble (completely)

density

0.786 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The structure of acetonitrile was determined by gas electron diffraction.[1]

Application

Acetonitrile is a high purity solvent used in electrical applications and in the synthesis of high-purity materials.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Structures of Hydrogen Cyanide and Acetonitrile as Studied by Gas Electron Diffraction
Karakida K, et al.
Bulletin of the Chemical Society of Japan, 47(2), 299-304 (1974)
Ranen Aviner et al.
PLoS genetics, 11(10), e1005554-e1005554 (2015-10-07)
Studying the complex relationship between transcription, translation and protein degradation is essential to our understanding of biological processes in health and disease. The limited correlations observed between mRNA and protein abundance suggest pervasive regulation of post-transcriptional steps and support the
Dwight R Stoll et al.
Analytical and bioanalytical chemistry, 407(1), 265-277 (2014-07-30)
In this paper, we describe the findings of a study aimed at assessing the detection sensitivity of comprehensive two-dimensional high-performance liquid chromatography (LCxLC) separation of a degraded active pharmaceutical ingredient (API) with UV absorption as the detection technique. Specifically, we
Seung-Ho Shin et al.
PloS one, 10(9), e0137011-e0137011 (2015-09-04)
Congenital cholesteatomas are epithelial lesions that present as an epithelial pearl behind an intact eardrum. Congenital and acquired cholesteatomas progress quite differently from each other and progress patterns can provide clues about the unique origin and pathogenesis of the abnormality.
Fang Xu et al.
The Plant cell, 27(5), 1547-1562 (2015-05-15)
Nod-like receptors (NLRs) serve as immune receptors in plants and animals. The stability of NLRs is tightly regulated, though its mechanism is not well understood. Here, we show the crucial impact of N-terminal acetylation on the turnover of one plant

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service