Merck
  • A novel miniaturized extraction capillary for determining gaseous formaldehyde by high-performance liquid chromatography.
A novel miniaturized extraction capillary for determining gaseous formaldehyde by high-performance liquid chromatography.
Analytical and bioanalytical chemistry (2014-09-10)
Ikuo Ueta, Suguru Mochizuki, Susumu Kawakubo, Tetsuo Kuwabara, Kiyokatsu Jinno, Yoshihiro Saito
ABSTRACT
A novel miniaturized sample extraction capillary was developed to provide a simple and sensitive method for analyzing gaseous formaldehyde (FA) using conventional high-performance liquid chromatography (HPLC). The extraction capillary was prepared by packing silica gel particles in a stainless steel capillary with a 1.6-mm o.d. Before the gaseous samples were collected, the silica gel particles were impregnated with a derivatizing reagent 2,4-dinitrophenylhydrazine (DNPH) by passing a solution of DNPH through the extraction capillary. FA was derivatized to form the corresponding hydrazone during the active sampling of a gaseous sample, and the derivative was sorbed by the silica gel. The capillary was then directly connected to a six-port valve, and desorption from the capillary and injection into the HPLC were achieved simultaneously by passing a solvent through the capillary. The run-to-run and device-to-device variabilities, as relative standard deviations, were both less than 6.9%. The limit of detection for FA was 2.5 ng/L using a sampling volume of 100 mL, and a sample collection time of 2 min. Acetaldehyde and acetone in a gaseous sample could also be simultaneously extracted and determined using this method. The applicability of the developed method for determining FA in real indoor air samples was confirmed by comparing the results obtained using the proposed method and a conventional sample preparation method. The developed method enables the rapid and simple sample preparation of gaseous FA and gives a good level of sensitivity.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Formaldehyde-2,4-DNPH Solution, certified reference material, TraceCERT®, 100 μg/mL in acetonitrile (as aldehyde), ampule of 5 × 1 mL
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Formaldehyde-2,4-dinitrophenylhydrazone, BCR®, certified reference material
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%