Merck
  • Structure elucidation and quantification of impurities formed between 6-aminocaproic acid and the excipients citric acid and sorbitol in an oral solution using high-resolution mass spectrometry and nuclear magnetic resonance spectroscopy.
Structure elucidation and quantification of impurities formed between 6-aminocaproic acid and the excipients citric acid and sorbitol in an oral solution using high-resolution mass spectrometry and nuclear magnetic resonance spectroscopy.
Journal of pharmaceutical and biomedical analysis (2015-02-04)
Anne Marie V Schou-Pedersen, Claus Cornett, Nils Nyberg, Jesper Østergaard, Steen Honoré Hansen
ABSTRACT
Concentrated solutions containing 6-aminocaproic acid and the excipients citric acid and sorbitol have been studied at temperatures of 50°C, 60°C, 70°C and 80°C as well as at 20°C. It has previously been reported that the commonly employed citric acid is a reactive excipient, and it is therefore important to thoroughly investigate a possible reaction between 6-aminocaproic acid and citric acid. The current study revealed the formation of 3-hydroxy-3,4-dicarboxy-butanamide-N-hexanoic acid between 6-aminocaproic acid and citric acid by high-resolution mass spectrometry (HRMS) and nuclear magnetic resonance spectroscopy (NMR). Less than 0.03% of 6-aminocaproic acid was converted to 3-hydroxy-3,4-dicarboxy-butanamide-N-hexanoic acid after 30 days of storage at 80°C. Degradation products of 6-aminocaproic acid were also observed after storage at the applied temperatures, e.g., dimer, trimer and cyclized 6-aminocaproic acid, i.e., caprolactam. No reaction products between D-sorbitol and 6-aminocaproic acid could be observed. 3-Hydroxy-3,4-dicarboxy-butanamide-N-hexanoic acid, dimer and caprolactam were also observed after storage at 20°C for 3 months. The findings imply that an oral solution of 6-aminocaproic acid is relatively stable at 20°C at the pH values 4.00 and 5.00 as suggested in the USP for oral formulations. Compliance with the ICH guideline Q3B is expected.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
ε-Caprolactam, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%