Skip to Content
Merck
All Photos(1)

Documents

33114

Sigma-Aldrich

Citric acid monohydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%

Synonym(s):

2-hydroxy-1,2,3-propanetricarboxylic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2 · H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
4018641
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

99.5-100.5% (calc. on anhydrous substance)
99.5-102%

form

solid

impurities

≤0.005% insolubles in water
≤2 ppm heavy metals (as Pb)
7.5-9.0% water (Karl Fischer)

ign. residue

≤0.01% (as SO4)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

anion traces

chloride (Cl-): ≤5 mg/kg
oxalate (C2O42-): ≤50 ppm
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤20 ppm

cation traces

Ca: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤3 ppm
Pb: ≤2 ppm
Zn: ≤5 mg/kg

suitability

complies for appearance of solution
complies for reaction against H2SO4

SMILES string

OC(CC(O)(C(O)=O)CC(O)=O)=O.O

InChI

1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2

InChI key

YASYEJJMZJALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Citric acid monohydrate is an organic acid. Its molar enthalpy of solution in water has been reported to be ΔsolHm (298.15K, m = 0.0203molkg-1) = (29061 ± 123)Jmol-1.

Application

Citric acid monohydrate may be used in the preparation of tris-citrate buffer and Otto I isolation buffer used in bioprotocols. It may also be used to improve the release of diltiazem hydrochloride from melt extruded matrix tablets.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

345.0 °F - closed cup

Flash Point(C)

173.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Citric acid monohydrate as a release-modifying agent in melt extruded matrix tablets.
Schilling SU, et al.
International Journal of Pharmaceutics, 361(1), 158-168 (2008)
Determination of the Developmental Origin of Seeds Containing Endosperm Using Flow Cytometric Analysis
Sailer C, et al.
PLoS Genetics, 5(11) (2015)
Methods of multilocus enzyme electrophoresis for bacterial population genetics and systematics.
R K Selander et al.
Applied and environmental microbiology, 51(5), 873-884 (1986-05-01)
Enthalpy of solution of oxalic, succinic, adipic, maleic, malic, tartaric, and citric acids, oxalic acid dihydrate, and citric acid monohydrate in water at 298.15K.
Apelblat A.
The Journal of Chemical Thermodynamics, 18(4), 351-357 (1986)
Soeren Lukassen et al.
The EMBO journal, 39(10), e105114-e105114 (2020-04-05)
The SARS-CoV-2 pandemic affecting the human respiratory system severely challenges public health and urgently demands for increasing our understanding of COVID-19 pathogenesis, especially host factors facilitating virus infection and replication. SARS-CoV-2 was reported to enter cells via binding to ACE2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service