Skip to Content
Merck
All Photos(5)

Key Documents

A2504

Sigma-Aldrich

6-Aminocaproic acid

≥99% (titration), powder

Synonym(s):

ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)5CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
906872
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥99% (titration)

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

mp

207-209 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

SMILES string

NCCCCCC(O)=O

InChI

1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)

InChI key

SLXKOJJOQWFEFD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

EACA is directly soluble in water at 25 mg/ml. As an inhibitor of plasmin it is has been utilized in the clotting buffer for fibrinogen assays. This buffer is 10 mM potassium and sodium phosphate, pH 6.4, with 0.20 g CaCl2, 5 g 6-Aminohexanoic acid, 1 g sodium azide, and 9 g NaCl in 1 liter. The buffer is stable indefinitely at room temperature.

Biochem/physiol Actions

EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator.
Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis.
Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 - 408.2 °F

Flash Point(C)

207 - 209 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuki Yamamoto et al.
Lab on a chip, 21(10), 1897-1907 (2021-05-20)
Engineered three-dimensional models of neuromuscular tissues are promising for use in mimicking their disorder states in vitro. Although several models have been developed, it is still challenging to mimic the physically separated structures of motor neurons (MNs) and skeletal muscle
Moamen Bydoun et al.
Molecular oncology, 12(11), 1895-1916 (2018-07-17)
Pancreatic cancer is arguably the deadliest cancer type. The efficacy of current therapies is often hindered by the inability to predict patient outcome. As such, the development of tools for early detection and risk prediction is key for improving outcome
Brian Hutton et al.
BMJ (Clinical research ed.), 345, e5798-e5798 (2012-09-13)
To estimate the relative risks of death, myocardial infarction, stroke, and renal failure or dysfunction between antifibrinolytics and no treatment following the suspension of aprotinin from the market in 2008 for safety reasons and its recent reintroduction in Europe and
María D Valls et al.
Frontiers in pharmacology, 12, 654104-654104 (2021-05-15)
Adenosine A2A receptor mediates the promotion of wound healing and revascularization of injured tissue, in healthy and animals with impaired wound healing, through a mechanism depending upon tissue plasminogen activator (tPA), a component of the fibrinolytic system. In order to
Sergio Guerrero-Castillo et al.
The EMBO journal, 40(21), e108648-e108648 (2021-09-21)
So-called ρ0 cells lack mitochondrial DNA and are therefore incapable of aerobic ATP synthesis. How cells adapt to survive ablation of oxidative phosphorylation remains poorly understood. Complexome profiling analysis of ρ0 cells covered 1,002 mitochondrial proteins and revealed changes in

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service