Skip to Content
Merck
All Photos(3)

Key Documents

S7547

Sigma-Aldrich

D-Sorbitol

≥98% (GC), BioXtra

Synonym(s):

D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

vapor density

<1 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

product line

BioXtra

Assay

≥98% (GC)

form

powder or chunks

impurities

<0.001% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

color

white

useful pH range

5.0-7.0 (25 °C, 182 g/L)

mp

98-100 °C (lit.)

solubility

H2O: 1 M, clear, colorless (room temperature)

anion traces

chloride (Cl-): <0.005%
sulfate (SO42-): <0.01%

cation traces

Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels. May be used to induce osmotic stress.

Biochem/physiol Actions

D-Sorbitol is a sugar alcohol that is commonly used as a sugar substitute. It occurs naturally and is also produced synthetically from glucose. The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement. D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals. Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoelectric focusing.
D E Garfin
Methods in enzymology, 182, 459-477 (1990-01-01)
Youngkook Kwon et al.
ChemSusChem, 6(3), 455-462 (2013-01-25)
This Full Paper addresses the electrocatalytic hydrogenation of glucose to sorbitol or 2-deoxysorbitol on solid metal electrodes in neutral media. Combining voltammetry and online product analysis with high-performance liquid chromatography (HPLC), provides both qualitative and quantitative information regarding the reaction
Anna Boccaccio et al.
Cellular and molecular life sciences : CMLS, 71(21), 4275-4283 (2014-04-29)
Two-pore channel proteins (TPC) encode intracellular ion channels in both animals and plants. In mammalian cells, the two isoforms (TPC1 and TPC2) localize to the endo-lysosomal compartment, whereas the plant TPC1 protein is targeted to the membrane surrounding the large
J M van Griensven et al.
Clinical pharmacology and therapeutics, 58(6), 631-640 (1995-12-01)
To examine the effect of diabetes mellitus on the pharmacokinetics of tolrestat and to investigate its effect on red blood cell sorbitol levels according to a new pharmacodynamic model for this class of drugs. Single and multiple doses of tolrestat
Beau Op de Beeck et al.
ChemSusChem, 6(1), 199-208 (2013-01-12)
The catalytic valorization of cellulose is currently subject of intense research. Isosorbide is among the most interesting products that can be formed from cellulose as it is a potential platform molecule and can be used for the synthesis of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service