Skip to Content
Merck
All Photos(1)

Key Documents

PHR1050

Supelco

Benzoic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39023903
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1055002

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

description

API family: benfluorex

API family

metronidazole, benfluorex, mefenamic acid

CofA

current certificate can be downloaded

autoignition temp.

1061 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

249 °C (lit.)

mp

121-125 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Benzoic acid may be used as a reference standard in determining the concentration of benzoic acid in food and milk samples using high performance liquid chromatography coupled with photodiode array detector (HPLC).

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB8257 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of benzoic and sorbic acids in Brazilian food
Tfouni.V.A.S and Toledo F.C.M
Food Control, 13, 117-123 (2002)
Determination of benzoic and sorbic acids in Brazilian food
Qi P, et al.
Food Control, 20, 414-418 (2009)
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.

Articles

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service