Skip to Content
Merck
All Photos(1)

Key Documents

24152

Supelco

Omegawax Capillary GC Column

L × I.D. 30 m × 0.32 mm, df 0.25 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32119290

material

fused silica

Agency

meets requirements for USP G16

packaging

pkg of 1 ea

parameter

50-280 °C temperature (isothermal or programmed)

Beta value

320

df

0.25 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.32 mm

matrix active group

Bonded; poly(ethylene glycol) phase

application(s)

agriculture
biofuels
clinical
food and beverages

column type

capillary polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: This column allows highly reproducible analyses of fatty acid methyl esters (FAMEs), specifically the omega 3 and omega 6 fatty acids. It is tested to ensure reproducible FAME equivalent chain length (ECL) values and resolution of key components.
USP Code: This column meets USP G16 requirements.
Phase:
  • Bonded
  • Poly(ethylene glycol)
Temp. Limits:
  • 50 °C to 280 °C (isothermal or programmed)

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Automated in-tube solid-phase microextraction coupled to high-performance liquid chromatography.
Ralf E and Pawliszyn J.
Analytical Chemistry, 69 (16), 3140-3147 (1997)
H Kataoka et al.
Analytical chemistry, 71(19), 4237-4244 (1999-10-12)
The technique of automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography/electrospray ionization mass spectrometry (LC/ESI-MS) was evaluated for the determination of beta-blockers in urine and serum samples. In-tube SPME is an extraction technique for organic compounds in aqueous samples
R O Adlof et al.
Lipids, 35(2), 131-135 (2000-04-11)
This paper deals with the reanalysis of serum lipids from previous studies in which deuterated fatty acids were administered to a single person. Samples were reanalyzed to determine if the deuterated fatty acids were converted to deuterium-labeled conjugated linoleic acid
Susana P Alves et al.
Journal of chromatography. A, 1216(26), 5130-5139 (2009-05-19)
Two gas-liquid chromatograph capillary columns for the analysis of fatty acids (FA) in ruminant fat are compared. Those columns are the CP-Sil 88 of 100 m long with a highly polar stationary phase and the Omegawax 250 of 30 m
M Vecka et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 770(1-2), 91-99 (2002-05-16)
Four capillary columns (A: CP-WAX 52 CB 25 m x 0.25 mm; B: CP WAX 52 CB 30 m x 0.25 mm; C: CP-WAX 58 CB 25 m x 0.25 mm, Chrompack; D: OMEGAWAX 320 30 m x 0.32 mm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service