Skip to Content
Merck
All Photos(2)

Key Documents

T6754

Sigma-Aldrich

Thymidine 3′:5′-cyclic monophosphate sodium salt

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2O7PNa
CAS Number:
Molecular Weight:
326.18
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

Assay

≥98%

form

solid

storage temp.

−20°C

SMILES string

[Na].CC1=CN(C2CC3OP(O)(=O)OCC3O2)C(=O)NC1=O

InChI

1S/C10H13N2O7P.Na.H/c1-5-3-12(10(14)11-9(5)13)8-2-6-7(18-8)4-17-20(15,16)19-6;;/h3,6-8H,2,4H2,1H3,(H,15,16)(H,11,13,14);;

InChI key

VPMQPENMZYNCDE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service