Skip to Content
Merck
All Photos(4)

Documents

D3876

Sigma-Aldrich

2′-Deoxyuridine 5′-monophosphate disodium salt

Sigma Grade

Synonym(s):

dUMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H11N2O8PNa2
CAS Number:
Molecular Weight:
352.15
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

grade

Sigma Grade

Assay

≥98% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(O)=O)N2C=CC(=O)NC2=O

InChI

1S/C9H13N2O8P.Na.H/c12-5-3-8(11-2-1-7(13)10-9(11)14)19-6(5)4-18-20(15,16)17;;/h1-2,5-6,8,12H,3-4H2,(H,10,13,14)(H2,15,16,17);;

InChI key

WXIVKKBDJOCRNB-UHFFFAOYSA-N

General description

2′-Deoxyuridine 5′-monophosphate disodium salt (dUMP) is a substrate for thymidylate synthase and is converted to deoxythymidine monophosphate (dTMP).

Application

2′-Deoxyuridine 5′-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.
2′-Deoxyuridine 5′-monophosphate disodium salt has been used:
  • in ultraperformance liquid chromatography-tandem mass spectrometry (UPLC/MS/MS) assay
  • to stimulate proliferation of peripheral blood mononuclear cell (PBMCs) exposed to influenza virus antigen
  • in thymidylate synthase activity assay Helicobacter pylori

Biochem/physiol Actions

2′-Deoxyuridine 5′-monophosphate disodium salt (dUMP) conversion to pyrimidine is inhibited by methotrexate. The inhibition of dUMP to deoxythymidine monophosphate (dTMP) at the methylation step is a key in controlling bacterial and eukaryotic cell growth.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary Newby et al.
Biochemistry, 45(24), 7415-7428 (2006-06-14)
The enzyme thymidylate synthase (TS) catalyzes the reductive methylation of 2'-deoxyuridine 5'-monophosphate (dUMP) to 2'-deoxythymidine 5'-monophosphate. Using kinetic and X-ray crystallography experiments, we have examined the role of the highly conserved Tyr-261 in the catalytic mechanism of TS. While Tyr-261
Dimitri Topalis et al.
The FEBS journal, 272(24), 6254-6265 (2005-12-13)
Anti-poxvirus therapies are currently limited to cidofovir [(S)-1-(3-hydroxy-2-phosphonylmethoxypropyl)cytosine], but drug-resistant strains have already been characterized. In the aim of finding a new target, the thymidylate (TMP) kinase from vaccinia virus, the prototype of Orthopoxvirus, has been overexpressed in Escherichia coli
Peter S Ludwig et al.
European journal of medicinal chemistry, 40(5), 494-504 (2005-05-17)
Amphiphilic anticancer prodrugs of 5'-fluoro-2'-deoxyuridine-5'-monophosphate (5-FdUMP) were synthesized according to the hydrogen phosphonate method by coupling lipophilic cytosine derivatives or a phospholipid with 5-fluoro-2'-deoxyuridine (5-FdU). Studies within the in vitro Anticancer Screen Program of the National Cancer Institute have demonstrated
Xiaoli Zhang et al.
Protein and peptide letters, 19(11), 1225-1230 (2012-04-20)
ThyX, a flavin-dependent thymidylate synthase that is involved in the synthesis of dTMP from dUMP, is a promising target for the development of novel antibacterial drugs that aimed at blocking the biosynthesis of dTMP, one of the building blocks of
Biochemistry: Anchors away.
Maria Paola Costi et al.
Nature, 458(7240), 840-841 (2009-04-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service