Skip to Content
Merck
All Photos(2)

Key Documents

C9625

Sigma-Aldrich

L-Carnosine

~99%

Synonym(s):

β-Alanyl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N4O3
CAS Number:
Molecular Weight:
226.23
Beilstein:
87671
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Carnosine, ~99%, crystalline

Assay

~99%

form

crystalline

color

white to off-white

mp

253 °C (dec.) (lit.)

application(s)

cell analysis

storage temp.

−20°C

SMILES string

NCCC(=O)N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)/t7-/m0/s1

InChI key

CQOVPNPJLQNMDC-ZETCQYMHSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-His

Application

L-Carnosine has been used to study its effects as an antioxidant on meat quality of pigs. It has also been used to study its effects on brain injury in a rat intracerebral hemorrhage model.

Biochem/physiol Actions

L-Carosine is a dipeptide found at millimolar concentration in brain, muscle and the lens of the eye. In model systems it is a potent antioxidant that scavenges oxygen free radicals and transition metal ions. It blocks protein-protein and protein-DNA cross-links induced by hypochlorite anions and toxic aldehydes such as acetaldehyde, formaldehyde, and malondialdehyde, the primary product of lipid peroxidation. It also inhibits nonenzymatic protein glycation induced by aldose and ketose reducing sugars and inhibits the formation of toxic advanced glycation end products (AGE). These activities make it of interest in studies of aging, atherosclerosis, Alzheimer′s disease, and the secondary effects of diabetes.
Dipeptide with potent antioxidant and antiglycation activity; blocks nonenzymatic glycosylation and protein cross-linking induced by reactive aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alexander A Boldyrev et al.
Physiological reviews, 93(4), 1803-1845 (2013-10-19)
Carnosine (β-alanyl-l-histidine) was discovered in 1900 as an abundant non-protein nitrogen-containing compound of meat. The dipeptide is not only found in skeletal muscle, but also in other excitable tissues. Most animals, except humans, also possess a methylated variant of carnosine
K N Roy Chengappa et al.
Schizophrenia research, 142(1-3), 145-152 (2012-10-27)
Targeting glutamatergic dysfunction provides an exciting opportunity to improve cognitive impairment in schizophrenia. One treatment approach has targeted inadequate antioxidant defenses at glutamatergic synapses. Animal and human data suggest NMDA antagonists worsen executive cognitive controls--e.g. increase perseverative responses and impair
Sanne Stegen et al.
PloS one, 10(3), e0121062-e0121062 (2015-03-25)
Muscle carnosine and its methylated form anserine are histidine-containing dipeptides. Both dipeptides have the ability to quench reactive carbonyl species and previous studies have shown that endogenous tissue levels are decreased in chronic diseases, such as diabetes. Rodent study: Skeletal
The Expression of Carnosine and Its Effect on the Antioxidant Capacity of Longissimus dorsi Muscle in Finishing Pigs Exposed to Constant Heat Stress
Peige Yang
Asian-Australasian Journal of Animal Sciences, 27(12), 1763-1772 (2014)
T L Dutka et al.
Journal of applied physiology (Bethesda, Md. : 1985), 112(5), 728-736 (2011-12-17)
There is considerable interest in potential ergogenic and therapeutic effects of increasing skeletal muscle carnosine content, although its effects on excitation-contraction (EC) coupling in human muscle have not been defined. Consequently, we sought to characterize what effects carnosine, at levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service