G1377
Gly-Gly-Gly
≥98% (TLC)
Synonym(s):
Glycyl-glycyl-glycine, Triglycine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH2CONHCH2CONHCH2COOH
CAS Number:
Molecular Weight:
189.17
Beilstein:
1711130
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
Gly-Gly-Gly,
Assay
≥98% (TLC)
Quality Level
form
powder
color
white
application(s)
peptide synthesis
SMILES string
NCC(=O)NCC(=O)NCC(O)=O
InChI
1S/C6H11N3O4/c7-1-4(10)8-2-5(11)9-3-6(12)13/h1-3,7H2,(H,8,10)(H,9,11)(H,12,13)
InChI key
XKUKSGPZAADMRA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Amino Acid Sequence
Gly-Gly-Gly
Application
Triglycine (Gly-Gly-Gly) is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator.
Biochem/physiol Actions
Substrate for reproducible serum protein measurements by the biuret reaction and for the assay of aminotripeptidases.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S J Woltman et al.
Analytical chemistry, 67(3), 541-551 (1995-02-01)
The reversible electrochemistry of the Cu(II)/Cu(III) couple was investigated for the copper(II) complexes of triglycine (G3), tetraglycine (G4), and pentaglycine (G5) in alkaline solution using a rotating ring-disk electrode (RRDE). The study was motivated by the need to elucidate electrochemical
Blake E Ziegler et al.
The journal of physical chemistry. A, 114(44), 11953-11963 (2010-10-26)
Hydrogen/deuterium exchange reactions involving protonated triglycine and deuterated ammonia (ND(3)) have been examined in the gas phase using a Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer. Ab initio and density functional theory (DFT) calculations have been carried out to
Sudipta Chakraborty et al.
Bioconjugate chemistry, 21(5), 969-978 (2010-04-15)
This report presents the synthesis and evaluation of (111)In(DOTA-6G-RGD(4)) (DOTA = 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetracetic acid; 6G-RGD(4) = E{G(3)-E[G(3)-c(RGDfK)](2)}(2) and G(3) = Gly-Gly-Gly), (111)In(DOTA-RGD(4)) (RGD(4) = E{E[c(RGDfK)](2)}(2)) and (111)In(DOTA-3G-RGD(2)) (3G-RGD(2) = G(3)-E[G(3)-c(RGDfK)](2)) as new radiotracers for imaging integrin alpha(v)beta(3)-positive tumors. The IC(50) values
Avisek Chatterjee et al.
The Journal of chemical physics, 129(10), 105104-105104 (2008-12-03)
X-ray photoelectron spectroscopy (XPS) has been used to investigate the core-level electronic structures of glycine (G) and its peptides, including glycyl-glycine (GG), diglycyl-glycine (GGG), and polyglycine (poly-G), in their powder forms. Increasing the number of G units in the peptides
Dipositively charged protonated a3 and a2 ions: generation by fragmentation of [La(GGG)(CH3CN)2]3+.
Tujin Shi et al.
Angewandte Chemie (International ed. in English), 47(43), 8288-8291 (2008-09-26)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service