Skip to Content
Merck
All Photos(1)

Key Documents

T3001

Sigma-Aldrich

(+)-α-Tocopherol acetate

oil or semi-solid, ~1360 IU/g

Synonym(s):

Vitamin E acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058007
PubChem Substance ID:
NACRES:
NA.79

biological source

plant

Quality Level

description

Synthesized from natural α-tocopherol

form

oil or semi-solid

specific activity

~1360 IU/g

color

yellow

mp

~25 °C (lit.)

storage temp.

room temp

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocopherol acetate is a fat soluble vitamin and a form of vitamin E. It can be obtained through dietary sources. RRRα-Tocopherol is a natural form of α-tocopherol.

Application

(+)-α-Tocopherol acetate has been used:
  • as a component of growth medium for retinal cell lines
  • as an external standard in high performance liquid chromatography (HPLC) to study its composition in leafy vegetables
  • in the preparation of trans-resveratrol-encapsulated lipid nanocarriers (R-nano)

Biochem/physiol Actions

α-Tocopherol is also known as a radical-chain breaker due to its anti-oxidant property.
Tocopherols (TCP) (vitamin E) are a series (α, β,γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Tocopherol acetate has properties similar but not identical to α-tocopherol.

Caution

Does not air oxidize.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resveratrol liposomes and lipid nanocarriers: comparison of characteristics and inducing browning of white adipocytes
Zu Y, et al.
Colloids and Surfaces. B, Biointerfaces, 164, 414-423 (2018)
Safety assessment of the substance alpha-tocopherol acetate for use in food contact materials
Bolognesi C, et al.
EFSA Journal, 14(3), 4412-4412 (2016)
Effects of natural (RRR α-tocopherol acetate) or synthetic (all-rac α-tocopherol acetate) vitamin E supplementation on reproductive efficiency in beef cows.
Horn M, Gunn P, Van Emon M, et al.
Journal of Animal Science, 88, 3121-3127 (2011)
Partial nutrient characterization of arugula (rocket-Eruca sativa L.) and the effect of heat treatment on its lipoxidase activity
Tassi EMM, et al.
Brazilian Journal of Food Technology, 21 (2018)
P C Allen et al.
Poultry science, 81(1), 41-48 (2002-03-12)
In two trials, broiler chickens, processed similarly to those placed in commercial operation, were fed, from 1 d of age, a range (13 to 200 ppm) of DL-alpha-tocopheryl acetate (VE-AC) levels, and the effects on the pathology of Eimeria maxima

Articles

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service