Skip to Content
Merck
All Photos(1)

Key Documents

T1157

Sigma-Aldrich

(+)-α-Tocopherol acetate

BioReagent, suitable for insect cell culture, ~1360 IU/g

Synonym(s):

Vitamin E acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058007
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

plant

Quality Level

description

Synthesized from natural α-tocopherol

product line

BioReagent

form

liquid (or semi-solid)

specific activity

~1360 IU/g

mol wt

Mw 472.74 g/mol

purified by

crystallization

technique(s)

cell culture | insect: suitable

color

white to yellow

solubility

chloroform: soluble 50 mg/mL, clear, colorless to yellow

storage temp.

room temp

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocopherol acetate is a semisynthetic acetate ester of naturally occurring α-tocopherol.

Application

(+)-α-Tocopherol acetate has been used:
  • as a supplement in the human embryonic kidney cells (HEK 293) to assess its impact on cell growth[1]
  • as a component of Dulbecco′s Modified Eagle Medium/Nutrient Mixture F-12 (DMEM/F12) for immortalized chondrocyte cell line[2]
  • as a component of serum-free medium for human colon tissue organ culture[3]
  • to test its antioxidant effects on cow articular chondrocytes[4]

Use in insect cell culture applications as an antioxidant.

Biochem/physiol Actions

α-Tocopherol acetate or vitamin E-acetate has antioxidant properties and reduces lipid peroxidation.[5] However, vitamin E acetate on pyrolysis leads to the generation of carcinogens like alkenes and benzenes.[6]

Caution

Since the M.P. is about 25 °C this preparation is usually an oil. Does not air oxidize.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael K Dame et al.
In vitro cellular & developmental biology. Animal, 47(1), 32-38 (2010-11-26)
We have recently shown that a multi-mineral extract from the marine red algae, Lithothamnion calcareum, suppresses colon polyp formation and inflammation in mice. In the present study, we used intact human colon tissue in organ culture to compare responses initiated
Horst Claassen et al.
Cell and tissue research, 319(3), 439-445 (2005-01-26)
Osteoarthritis (OA) is aggravated in menopausal women possibly because of changed serum estrogen levels. Estradiol has been postulated to affect oxidative stress induced by reactive oxygen species (ROS) in articular chondrocytes. We generated ROS in cultured bovine articular chondrocytes by
Role of vitamin E-acetate on cisplatin induced genotoxicity: An in vivo analysis
Mazumdar M, et al.,
Central European Journal of Biology, 7(2), 334-342 (2012)
Optimization of HEK 293 cell growth by addition of non-animal derived components using design of experiments.
Laura Cervera et al.
BMC proceedings, 5 Suppl 8, P126-P126 (2012-03-01)
Michael K Dame et al.
In vitro cellular & developmental biology. Animal, 46(2), 114-122 (2009-11-17)
Normal and neoplastic human colon tissue obtained at surgery was used to establish conditions for organ culture. Optimal conditions included an atmosphere of 5% CO2 and 95% O2; tissue partially submerged with mucosa at the gas interface; and serum-free medium

Articles

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

Importance and uses of vitamin E (tocopherol) in serum-free eukaryotic, including hybridoma and chinese hamster ovary (CHO), cell cultures.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service