Skip to Content
Merck
All Photos(3)

Documents

T3376

Sigma-Aldrich

DL-α-Tocopherol acetate

≥96% (HPLC)

Synonym(s):

Vitamin E acetate, all-rac-α-Tocopheryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
Beilstein:
97512
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Assay

≥96% (HPLC)

form

viscous liquid

technique(s)

HPLC: suitable

color

colorless to dark yellow

density

0.96 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocopherol is a predominant form of vitamin E in human and animal tissues and also in leaf chloroplasts. It is the major form in supplements. DL-α-Tocopherol acetate is considered as the only storage form of vitamin E.

Application

DL-α-Tocopherol acetate has been used to study it effect on blood pressure and lipidic profile in streptozotocin-induced diabetes mellitus rats. It has also been used as a standard in α-tocopherol analysis using plasma sample and mature fresh tobacco leaves.

Biochem/physiol Actions

Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Tocopherol acetate has properties similar but not identical to α-tocopherol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High levels of dietary unsaturated fat decrease alpha-tocopherol content of whole body, liver, and plasma of chickens without variations in intestinal apparent absorption
Villaverde C, et al.
Poultry Science, 87(3), 497-505 (2008)
Nutrient Requirements of Fish and Shrimp (2011)
Lowered plasma vitamin C, but not vitamin E, concentrations in dementia patients
Charlton KE, et al.
The Journal of Nutrition, Health & Aging, 8(2), 99-108 (2004)
gamma-Tocopherol, the major form of vitamin E in the US diet, deserves more attention
Jiang Q, et al.
American Journal of Clinical Nutrition, 74(6), 714-722 (2001)
Effect of alpha-tocopherol supplementation on blood pressure and lipidic profile in streptozotocin-induced diabetes mellitus in spontaneously hypertensive rats
da Costa VAV and Vianna LM
Clinica Chimica Acta; International Journal of Clinical Chemistry, 351(1-2), 101-104 (2005)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service