Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
G6013
L-Glutathione reduced
≥98.0%, suitable for cell culture, BioReagent
Synonym(s):
Glutathione, Reduced, γ-L-Glutamyl-L-cysteinyl-glycine, GSH
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
product line
BioReagent
Quality Level
Assay
≥98.0%
form
powder
technique(s)
cell culture | mammalian: suitable
color
white
mp
192-195 °C (dec.) (lit.)
solubility
H2O: 50 mg/mL
storage temp.
2-8°C
SMILES string
N[C@@H](CCC(=O)N[C@@H](CS)C(=O)NCC(O)=O)C(O)=O
InChI
1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1
InChI key
RWSXRVCMGQZWBV-WDSKDSINSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Amino Acid Sequence
Application
Biochem/physiol Actions
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn how glutathione and other components affect serum-free cell culture systems for protein biomanufacturing.
Learn how glutathione and other components affect serum-free cell culture systems for protein biomanufacturing.
Learn how glutathione and other components affect serum-free cell culture systems for protein biomanufacturing.
Learn how glutathione and other components affect serum-free cell culture systems for protein biomanufacturing.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
How does the storage temperature relate to shipping conditions?
1 answer-
The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What is the GSH level in plasma?
1 answer-
It is in the micromolar range (approximately 4.5 mM). Chawla RK, Lewis FW, Kutner MH, et al. Plasma cysteine, cysteine, and glutathione in cirrhosis. Gastroenterology, 87, 770-776 (1984).
Helpful?
-
-
What are the GSH levels in human tissues?
1 answer-
The GSH levels in human tissues normally range from 0.1 to 10 mM, most concentrated in the liver (up to 10 mM) and in the spleen, kidney, lens, erythrocytes, and leukocytes. Bremer HJ. Duran M, Kamerling JP, et al. Glutathione. In: Bremer HJ,DuranM, Kamerling JP, et al, eds. Distubance of Amino Acid Metabolism: Clinical Chemistry and Diagnosis. Baltimore-Munich: Urban and Schwarzenber; 1981:80-82.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service