Skip to Content
Merck
All Photos(2)

Documents

T2786

Sigma-Aldrich

Tungstosilicic acid hydrate

Synonym(s):

Silicotungstic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H4[Si(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2878.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

form

powder

solubility

water: 100 mg/mL, clear, colorless

SMILES string

O.O[Si](O)(O)O.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]2(=O)O[W](=O)(=O)O[W](=O)(=O)O2.O=[W]3(=O)O[W](=O)(=O)O[W](=O)(=O)O3.O=[W]4(=O)O[W](=O)(=O)O[W](=O)(=O)O4

InChI

1S/H4O4Si.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h1-4H;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

ACTPFSYIGLFGLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tungstosilicic acid hydrate is widely used as a catalyst in esterification, oxidation, and other chemical reactions.

Application

Reactant for:
  • Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid material
  • Synthesis of SnII-substituted Keggin-type polyoxometalates as photocatalysts
  • Preparation of silicotungstic acid/organically modified silane proton-conducting membranes
  • Preparation of charge transfer compounds with Keggin anions
  • Preparation of catalyst for catalytic liquid-phase oxidation of xylene
Tungstosilicic acid can be used as a reactant for:
  • Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid material.
  • Synthesis of Sn(II)-substituted Keggin-type polyoxometalates as photocatalysts.
  • Preparation of silicotungstic acid/organically modified silane proton-conducting membranes.
  • Preparation of charge transfer compounds with Keggin anions.
  • Preparation of catalyst for catalytic liquid-phase oxidation of xylene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogenization of a Tungstosilicic Acid Catalyst for Esterification of Bio-Oil Model Compound
P Prasertpong, et al.
Catalysts, 13, 38-38 (2023)
Silicotungstic acid supported zirconia: An effective catalyst for esterification reaction
KM Parida, et al.
Journal of Molecular Catalysis. B, Enzymatic, 275, 77-83 (2007)
Synthesis of proton conducting tungstosilicate mesoporous materials and polymer composite membranes.
Feng F, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 81(1-3), 217-234 (2005)
Catalytic liquid phase oxidation of p-xylene using transition metal substituted polyoxometalates.
Alekar N A, et al.
Indian Journal of Chemistry, 39A, 439-441 (2000)
Synthesis and characterization of hybrid molecular material prepared by ionic liquid and silicotungstic acid.
Rajkumar T and Rao G R
Materials Chemistry and Physics, 112(3), 853-857 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service