Skip to Content
Merck
All Photos(3)

Key Documents

419028

Sigma-Aldrich

Cellulose acetate

average Mn ~50,000 by GPC

Synonym(s):

Acetylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

mol wt

average Mn ~50,000 by GPC

extent of labeling

39.7 wt. % acetyl

impurities

≤3.0% water

refractive index

n20/D 1.475 (lit.)

density

1.3 g/mL at 25 °C (lit.)

SMILES string

O1[C@H](C(C(C(C1COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)O[C@@H]2C(OC(C(C2OC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C

InChI

1S/C28H38O19/c1-11(29)37-9-19-21(39-13(3)31)23(40-14(4)32)26(43-17(7)35)28(46-19)47-22-20(10-38-12(2)30)45-27(44-18(8)36)25(42-16(6)34)24(22)41-15(5)33/h19-28H,9-10H2,1-8H3/t19?,20?,21?,22-,23?,24?,25?,26?,27?,28+/m1/s1

InChI key

WOTQVEKSRLZRSX-OLIDUAMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose acetate (CA) is a class of cellulose based esters which can be produced by a reaction of cellulose with acetic acid and acetic anhydride. It can be used in a variety of forms such as films, membranes, and fibers. It can be used as a biodegradable polymer in medical and automotive applications.[1]

Application

CA can be used as a material in packaging, clothing and pharmaceuticals industries.[2] It can also be used in the formation of polymeric electrolyte for the fabrication of electrochemical devices like fuel cells, solar cells and supercapacitors.[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of bio polymer electrolyte based on cellulose acetate-ammonium nitrate for potential use in electrochemical devices
Monisha S, et al.
Carbohydrate Polymers, 157(4), 38-47 (2017)
Properties and applications of cellulose acetate
Macromolecular Symposia, 262(1), 89-96 (2008)
Additive manufacturing of cellulosic materials with robust mechanics and antimicrobial functionality
Pattinson SW and Hart AJ
Advanced Materials Technologies, 2(4), 1600084-1600084 (2017)
Rocktotpal Konwarh et al.
Carbohydrate polymers, 92(2), 1100-1106 (2013-02-13)
This work focuses on the use of statistical approach in optimizing shape-size accord of electrospun cellulose acetate (CA) mats - an apt material for biomedical and industrial applications. Modulation of three processing parameters, namely potential difference, distance between tip-to-collector and
Deng-Guang Yu et al.
Carbohydrate polymers, 90(2), 1016-1023 (2012-07-31)
This study investigates the use of a modified coaxial electrospinning process in the production of drug-loaded cellulose acetate (CA) nanofibers. With CA employed as a filament-forming matrix and ketoprofen (KET) as an active pharmaceutical ingredient, modified coaxial processes using sheath

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service