Skip to Content
Merck
All Photos(1)

Key Documents

T2751

Sigma-Aldrich

D-(−)-Tagatose

≥98% (HPLC)

Synonym(s):

D-lyxo-2-Hexulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724555
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98% (HPLC)

form

powder or crystals

optical activity

[α]25/D -6.5 to -5.0 °, c = 1% (w/v) in water

sweetness

0.9 × sucrose

technique(s)

HPLC: suitable

color

white

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5+,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-PQLUHFTBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(-)-Tagatose has been used as a carbohydrates for fermentation. It has also been used as one of the standards to confirm the identity of majority of the metabolites selected by least absolute shrinkage and selection operator (LASSO).

Biochem/physiol Actions

D-Tagatose, a ketohexose acts as a low-calorie functional sweetener. Tagatose can be used as a preservative in cosmetic, detergent and pharmaceutical formulations.Tagatose is also used in diet soft drinks, chewing gum, frozen yogurt and non-fat ice cream.
Potential sugar substitute rarely found in nature. Produced using a biotransformation method with L-arabinose isomerase as the biocatalyst and D-galactose as the substrate.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbohydrate Basics: Sugars, Starches and Fibers in Foods and Health: Healthy Carbohydrate Choices, Roles and Applications in Nutrition, Food Science and the Culinary Arts
Marcus JB
Culture, Health & Sexuality, 149-187 (2013)
Biochemical Characterization of Heat-Tolerant Recombinant l-Arabinose Isomerase from Enterococcus faecium DBFIQ E36 Strain with Feasible Applications in d-Tagatose Production
Manzo RM, et al.
Molecular Biotechnology, 61(6), 385-399 (2019)
Recommended test panel for differentiation of Klebsiella species on the basis of a trilateral interlaboratory evaluation of 18 biochemical tests
Hansen DS, et al.
Journal of Clinical Microbiology, 42(8), 3665-3669 (2004)
Metabolomic characterization of hepatocellular carcinoma in patients with liver cirrhosis for biomarker discovery
Di PC, et al.
Cancer Epidemiology, Biomarkers & Prevention, 26(5), 675-683 (2017)
Ji Hoon Koh et al.
Food microbiology, 36(1), 7-13 (2013-06-15)
Synbiotics, the combination of prebiotics and probiotics, has been shown to produce synergistic effects that promote gastrointestinal well-being of host. Tagatose is a low calorie food ingredient with putative health-promoting benefits. Herein, we investigated its synbiotic impact on the viability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service