Skip to Content
Merck
All Photos(1)

Key Documents

220477

Sigma-Aldrich

D-(−)-Lyxose

99%

Synonym(s):

D-Lyxopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723083
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]25/D -15.8°, c = 4 in H2O

mp

108-112 °C (lit.)

SMILES string

O[C@@H]1COC(O)[C@@H](O)[C@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4+,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-AGQMPKSLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judit E Sponer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(32), 9990-9998 (2008-09-24)
Recent experimental studies suggest that complexation with borate minerals stabilizes ribose, and that the borate complex of ribose is more stable than those of related aldopentoses, that is, arabinose, lyxose, and xylose. These findings have revived the debate on the
Laura K Jennings et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(36), 11353-11358 (2015-08-28)
Biofilm formation is a complex, ordered process. In the opportunistic pathogen Pseudomonas aeruginosa, Psl and Pel exopolysaccharides and extracellular DNA (eDNA) serve as structural components of the biofilm matrix. Despite intensive study, Pel's chemical structure and spatial localization within mature
M Jørgensen et al.
The Journal of organic chemistry, 66(13), 4625-4629 (2001-06-26)
The combination of a Wittig olefination and a dihydroxylation reaction constitutes a facile synthetic protocol for the transformation of unprotected carbohydrates into higher sugars. The Wittig reaction is carried out with tert-butyl or diphenylmethyl ester stabilized phosphoranes to give (E)-configured
Michal Letek et al.
PLoS genetics, 6(9), e1001145-e1001145 (2010-10-14)
We report the genome of the facultative intracellular parasite Rhodococcus equi, the only animal pathogen within the biotechnologically important actinobacterial genus Rhodococcus. The 5.0-Mb R. equi 103S genome is significantly smaller than those of environmental rhodococci. This is due to
Carsten Hofmann et al.
Chemistry & biology, 12(10), 1137-1143 (2005-10-26)
The oligosaccharide antibiotic avilamycin A is composed of a polyketide-derived dichloroisoeverninic acid moiety attached to a heptasaccharide chain consisting of six hexoses and one unusual pentose moiety. We describe the generation of mutant strains of the avilamycin producer defective in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service