Skip to Content
Merck
All Photos(3)

Key Documents

P8043

Sigma-Aldrich

D-Psicose

≥95% (HPLC)

Synonym(s):

(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, D-ribo-2-Hexulose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

solid

impurities

≤25% water (Karl Fischer)

solubility

water: 100 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@@H](O)[C@@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5+,6-/m0/s1

InChI key

BJHIKXHVCXFQLS-LFRDXLMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Psicose protects and preserves pancreatic β-islets through the maintenance of hyperglycemia and by the prevention of fat accumulation in Otsuka Long-Evans Tokushima Fatty rats.

Application

D-Psicose has been used in a study to assess the bioconversion of D-fructose to D-allose. It has also been used in a study that investigated the characteristics and antioxidant activity of maillard reaction products.

Other Notes

Mixed anomers
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid S Surono et al.
Nutrition & metabolism, 17, 77-77 (2020-09-25)
The gut microbiota has been shown to be involved in the development and severity of type 2 diabetes. The aim of the present study was to test the effect of 4-week functional food ingredient feeding, alone or in combination, on
Akram Hossain et al.
Biochemical and biophysical research communications, 425(4), 717-723 (2012-08-11)
Rare sugar D-psicose has cropped up as a non-toxic and effective compound to protect and preserve pancreatic β-islets in the growing type 2 diabetes mellitus (T2DM) rats through the regulation of glucose and fat metabolism. The present study was undertaken
S H Baek et al.
Journal of food science, 75(2), H49-H53 (2010-05-25)
D-psicose has been implicated in glycemic control in recent animal and human studies. In this study, the effects of D-psicose on glycemic responses, insulin release, and lipid profiles were compared with those of D-glucose and D-fructose in a genetic diabetes
Bioconversion of D-fructose to D-allose by novel isomerases
Bai, W., et al.
Sheng Wu Gong Cheng Xue Bao (Chinese journal of biotechnology), 28, 28-28 (2012)
Wanmeng Mu et al.
Applied microbiology and biotechnology, 94(6), 1461-1467 (2012-05-10)
D-Psicose is a hexoketose monosaccharide sweetener, which is a C-3 epimer of D-fructose and is rarely found in nature. It has 70 % relative sweetness but 0.3 % energy of sucrose, and is suggested as an ideal sucrose substitute for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service