Skip to Content
Merck
All Photos(1)

Key Documents

1236801

USP

Epilactose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-O-β-D-Galactopyranosyl-D-mannose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

epilactose

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@H](O)C=O)[C@H](O)CO

InChI

1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5-,6-,7-,8+,9+,10-,11-,12+/m1/s1

InChI key

DKXNBNKWCZZMJT-QMRWEYQWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Epilactose USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as:
  • Lactulose Concentrate
  • Lactulose Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei Li et al.
Biochemistry, 47(1), 378-387 (2007-12-15)
The wbsJ gene from Escherichia coli O128:B12 encodes an alpha1,2-fucosyltransferase responsible for adding a fucose onto the galactose residue of the O-antigen repeating unit via an alpha1,2 linkage. The wbsJ gene was overexpressed in E. coli BL21 (DE3) as a
E H Reimerdes et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 181(5), 408-411 (1985-11-01)
A method is described for the analysis of carbohydrates in milk and milk products which develop during heat treatment. It is based on the separation of their borate complexes by anion exchange chromatography using two optimized buffer systems. A reliable
Hiroki Sato et al.
Bioscience, biotechnology, and biochemistry, 76(8), 1584-1587 (2012-08-11)
Cellobiose 2-epimerase (CE) efficiently forms epilactose which has several beneficial biological functions. A thermostable CE from Rhodothermus marinus was immobilized on Duolite A568 and packed into a column. Lactose (100 g/L) was supplied to the reactor, kept at 50 °C
J Watanabe et al.
Journal of dairy science, 91(12), 4518-4526 (2008-11-29)
We recently reported that cellobiose 2-epimerase from Ruminococcus albus effectively converted lactose to epilactose. In this study, we examined the biological effects of epilactose on intestinal microbiota, bile acid metabolism, and postadministrative plasma glucose by animal tests. Dietary supplementation with
Mariko Miyasato et al.
Bioscience, biotechnology, and biochemistry, 68(10), 2086-2090 (2004-10-27)
The regioselectivity of beta-galactosidase derived from Bacillus circulans ATCC 31382 (beta-1,3-galactosidase) in transgalactosylation reactions using D-mannose as an acceptor was investigated. This D-mannose associated regioselectivity was found to be different from reactions using either GlcNAc or GalNAc as acceptors, not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service