Skip to Content
Merck
All Photos(1)

Key Documents

T1660

Sigma-Aldrich

Nε,Nε,Nε-Trimethyllysine hydrochloride

≥97% (TLC)

Synonym(s):

1-Pentanaminium, 5-amino-5-carboxy-N,N,N-trimethyl-, chloride (1:1), (5S)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20N2O2 · HCl
CAS Number:
Molecular Weight:
224.73
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Assay

≥97% (TLC)

form

powder

contains

salts and water as balance

composition

Amino acid content, ~75%

technique(s)

LC/MS: suitable

color

white

storage temp.

−20°C

SMILES string

[Cl-].C[N+](C)(C)CCCC[C@H](N)C(O)=O

InChI

1S/C9H20N2O2.ClH/c1-11(2,3)7-5-4-6-8(10)9(12)13;/h8H,4-7,10H2,1-3H3;1H/t8-;/m0./s1

InChI key

ZKIJKCHCMFGMCM-QRPNPIFTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naomi van Vlies et al.
Analytical biochemistry, 354(1), 132-139 (2006-05-19)
Although the mouse frequently is used to study metabolism and deficiencies therein, little is known about carnitine biosynthesis in this animal. To this point, only laborious procedures have been described to measure the activity of carnitine biosynthesis enzymes using subcellular
Jean-François Couture et al.
The Journal of biological chemistry, 281(28), 19280-19287 (2006-05-10)
SET domain enzymes represent a distinct family of protein lysine methyltransferases in eukaryotes. Recent studies have yielded significant insights into the structural basis of substrate recognition and the product specificities of these enzymes. However, the mechanism by which SET domain
Lynnette M A Dirk et al.
Biochemistry, 46(12), 3905-3915 (2007-03-07)
Processive versus distributive methyl group transfer was assessed for pea Rubisco large subunit methyltransferase, a SET domain protein lysine methyltransferase catalyzing the formation of trimethyllysine-14 in the large subunit of Rubisco. Catalytically competent complexes between an immobilized form of des(methyl)
Zhenyu Lu et al.
Journal of the American Chemical Society, 131(41), 14928-14931 (2009-10-01)
Histone modifications are implicated in epigenetic inheritance and are of central importance in regulating chromatin structure and gene expression. A prototype example is the trimethylation (Me3) of lysine 9 on histone 3 (H3), which is a readout by an aromatic
Frank Hirche et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(22), 2158-2162 (2009-06-16)
For the investigation of the metabolism and biosynthesis of carnitine, sensitive determination of carnitine and its metabolic precursors, trimethyllysine and gamma-butyrobetaine, is required. We present here a new simplified method for the analysis of carnitine, its acetyl- and propyl esters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service