Skip to Content
Merck
All Photos(1)

Key Documents

26978

Sigma-Aldrich

Choline chloride

BioUltra, ≥99.0% (AT)

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
Beilstein:
3563126
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
powder or crystals
solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.

biological source

synthetic

grade

reagent grade

product line

BioUltra

Assay

≥99.0% (AT)

form

powder or crystals

impurities

insoluble matter, passes filter test

color

colorless to white

pH

5.0-7.5 (25 °C, 1 M in H2O)

mp

302-305 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.015
λ: 280 nm Amax: 0.010

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Choline is an essential nutrient, commonly grouped with the B complex vitamins, that plays key roles in many biological processes.

Substrates

Choline acetyltransferase substrate

Other Notes

Restores cholinergic, biochemical and pharmacological activities in striata of decorticated rats[1]
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Consolo et al.
Journal of neurochemistry, 54(2), 571-577 (1990-02-01)
Interruption of the corticostriatal pathway by undercutting the frontal cortex resulted after 2 weeks in a 40% reduction of basal acetylcholine (ACh) release in vivo, and in inhibition of the striatal sodium-dependent high-affinity uptake of choline (SDHACU) to the same
Elisabeth Traiffort et al.
Molecular aspects of medicine, 34(2-3), 646-654 (2013-03-20)
The Na(+)-independent, high affinity choline carrier system proposed to supply choline for the synthesis of cell membrane phospholipids was recently associated with SLC44 family members (SLC44A1-5) also called choline-like transporter family. SLC44A1 is widely expressed throughout the nervous system in
Mohsen Beheshti et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 833-840 (2013-04-06)
We evaluated the potential of (18)F-fluoromethyldimethyl-2-hydroxyethyl-ammonium (FCH) PET/CT in the detection of recurrent disease or distant metastases and correlated its diagnostic accuracy with prostate-specific antigen (PSA) levels in prostate cancer patients with biochemical evidence of recurrence. Furthermore, the influences of
Robert A Koeth et al.
Nature medicine, 19(5), 576-585 (2013-04-09)
Intestinal microbiota metabolism of choline and phosphatidylcholine produces trimethylamine (TMA), which is further metabolized to a proatherogenic species, trimethylamine-N-oxide (TMAO). We demonstrate here that metabolism by intestinal microbiota of dietary L-carnitine, a trimethylamine abundant in red meat, also produces TMAO
Sotirios Chondrogiannis et al.
European journal of nuclear medicine and molecular imaging, 40(9), 1356-1364 (2013-05-15)
The aims of the study were (a) to evaluate the diagnostic role, by means of positive detection rate (PDR), of ¹⁸F-choline (CH) positron emission tomography (PET)/CT in patients with prostate cancer treated with radiotherapy, with curative intent, and suspicion of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service