Skip to Content
Merck
All Photos(1)

Key Documents

67520

Sigma-Aldrich

1-Methyl-L-histidine

≥98.0% (TLC), for peptide synthesis

Synonym(s):

3-(1-Methylimidazol-4-yl)-L-alanine, 3-Methyl-L-histidine (archaic), Nτ-Methyl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3O2
CAS Number:
Molecular Weight:
169.18
Beilstein:
9727
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

1-Methyl-L-histidine, ≥98.0% (TLC)

Quality Level

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]20/D −24±1°, c = 2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

impurities

≤5% water

mp

~240 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cn1cnc(C[C@H](N)C(O)=O)c1

InChI

1S/C7H11N3O2/c1-10-3-5(9-4-10)2-6(8)7(11)12/h3-4,6H,2,8H2,1H3,(H,11,12)/t6-/m0/s1

InChI key

BRMWTNUJHUMWMS-LURJTMIESA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Natural but non-proteinogenic amino acid; employed as index of muscle protein breakdown

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Sjölin et al.
Metabolism: clinical and experimental, 38(1), 23-29 (1989-01-01)
The present investigation was undertaken in order to determine the release of 3-methylhistidine (3MH) from the splanchnic region and from the leg, and the contributions these make to the increase in urinary 3MH excretion in infection. Thirteen febrile patients with
3-Methylhistidine as a measure of skeletal muscle protein breakdown in human subjects: the case for its continued use.
F J Ballard et al.
Clinical science (London, England : 1979), 65(3), 209-215 (1983-09-01)
P C Colombani et al.
International journal of sport nutrition, 9(2), 181-201 (1999-06-11)
A field study was performed to investigate the acute influence of a milk protein hydrolysate supplemented drink (CHO+PRO) on metabolism during and after a marathon run compared to the same drink without protein (CHO). Carbohydrate metabolites and hormones were not
V.R. Young et al.
The Journal of Biological Chemistry, 247, 293-293 (1972)
J Sjölin et al.
Metabolism: clinical and experimental, 36(12), 1175-1184 (1987-12-01)
In order to investigate whether the urinary excretion of 1-methylhistidine (1MH) might serve as an objective indicator of meat ingestion and exogenous 3-methylhistidine (3MH) intake, healthy subjects were fed an ovolactovegetarian diet. At five-day intervals they were given meat of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service