Skip to Content
Merck
All Photos(1)

Documents

72332

Supelco

Nonadecanoic acid

analytical standard

Synonym(s):

C19:0, NSC 11914, Nonadecylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17COOH
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786261
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

68-70 °C (lit.)

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C19H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21/h2-18H2,1H3,(H,20,21)

InChI key

ISYWECDDZWTKFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonadecanoic acid has been used as an internal standard for the quantitative determination of acid catalyzed methylation of fatty acid methyl esters in microalgae, Selenastrum sp. in the presence of bacteria, using gas chromatography coupled with mass spectrometry.(GC-MS).
Nonadecanoic acid may be used as an analytical reference standard for the quantification of the analyte in Notopterygium forbesii Boiss using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Culturing of Selenastrum on diluted composting fluids; conversion of waste to valuable algal biomass in presence of bacteria
Marika T, et al.
BioResources, 238, 205-213 (2017)
Ahmad Al-Mrabeh et al.
Cell metabolism, 31(2), 233-249 (2019-12-24)
The role of hepatic lipoprotein metabolism in diet-induced remission of type 2 diabetes is currently unclear. Here, we determined the contributions of hepatic VLDL1-triglyceride production rate and VLDL1-palmitic acid content to changes in intra-pancreatic fat and return of first phase
Bruno Levecke et al.
Parasites & vectors, 7, 255-255 (2014-06-03)
Cysteine proteinases (CPs) from papaya (Carica papaya) possess anthelmintic properties against human soil-transmitted helminths (STH, Ascaris lumbricoides, Trichuris trichiura and hookworm), but there is a lack of supportive and up-to-date efficacy data. We therefore conducted two randomized controlled trials in
Bhushan V Kulkarni et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(12), R879-R885 (2014-04-04)
Lingual lipase generates nonesterified fatty acids (NEFA) from dietary fats during oral processing by lipolysis. Lingual lipase in rodents has strong lipolytic activity and plays a critical role in oral detection of fats. The functional activity of lingual lipase during
Shijuan Zhang et al.
Talanta, 98, 95-100 (2012-09-04)
A new labeling reagent for fatty acids, 1-(9H-carbazol-9-yl) propan-2-yl-methanesulfonate (CPMS), has been synthesized and successfully applied to the HPLC determination of fatty acids in traditional Chinese herb Notopterygium forbesii Boiss. The reaction of CPMS with fatty acids could proceed easily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service