Skip to Content
Merck
All Photos(4)

Documents

H3500

Sigma-Aldrich

Heptadecanoic acid

≥98%

Synonym(s):

C17:0, Heptadecylic acid, Margaric acid, NSC 3743

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15COOH
CAS Number:
Molecular Weight:
270.45
Beilstein:
1781004
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98%

form

powder

bp

227 °C/100 mmHg (lit.)

mp

59-61 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C17H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3,(H,18,19)

InChI key

KEMQGTRYUADPNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Margaric acid (Heptadecanoic acid, C17) may be used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Gyun Lim et al.
Korean journal for food science of animal resources, 34(3), 287-296 (2014-01-01)
The physicochemical and functional traits for loin muscles of Hanwoo steers were compared by quality grade (QG). A total of 500 Hanwoo steers were slaughtered, their carcasses were categorized into four groups (QG 1++, 1+, 1, and 2), and the
Louwrens C Hoffman et al.
Foods (Basel, Switzerland), 9(5) (2020-05-21)
The common warthog (Phacochoerus africanus) has historically been hunted and consumed by rural communities throughout its distribution range in Africa. This study aims to develop a processed product from warthog meat in the form of back bacon (Longissimus thoracis et
Sheetal Saini et al.
Frontiers in cellular and infection microbiology, 10, 406-406 (2020-08-28)
Visceral leishmaniasis is a neglected tropical disease caused by Leishmania (L.) donovani parasite in the Indian subcontinent. Macrophages (mϕ) are the harboring cells for parasite and their interactions dictate the pathogenesis of this disease. Polyunsaturated fatty acids are an integral
L C Hoffman et al.
Meat science, 93(3), 696-702 (2012-12-19)
Nutritional value of Dorper (n=10) and Merino (n=10) by-products were evaluated. Proximate composition differed between organs and breeds with Merino heart (68.9 g/100 g), spleen (77.2 g/100 g) and testicles (83.7 g/100 g) having higher moisture contents than their Dorper
Sara Wilhelmina Erasmus et al.
Foods (Basel, Switzerland), 9(8) (2020-08-17)
Alternative protein sources are gaining increasing global attention as a solution to address future protein demands. Determining the chemical composition of meat alternatives is vital to confirm that it is nutritious, but also to increase product value and promote its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service