SRP0324
MECP2 human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
MRX16, PPMX, RTS, methyl CpG binding protein 2 (Rett syndrome)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
36 kDa
packaging
pkg of 50 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MECP2(4204)
General description
Human recombinant Methyl CpG binding Protein 2 (Rett syndrome) (MECP2), (GenBank Accession No. NM_004992), amino acids 78-162 with N-terminal GST-tag, MW= 36 kDa, expressed in an E. coli expression system.
Methyl-CpG binding protein 2 (MECP2) is encoded by the gene mapped to human chromosome Xq28. Higher molecular weight form of hMeCP2 is expressed in frontal cortex nuclear and synaptic fractions, and in lymphoid cells. Fibroblast and lymphoblastoid strains of female patients with Rett syndrome and MeCP2 transfected cells also show expression of high molecular weight form of MeCP2.2The encoded chromosomal protein is characterized with the 80 amino acid-containing methyl-CpG binding domain, involved in chromosomal localization of the protein.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Biochem/physiol Actions
Methyl-CpG binding protein 2 (MECP2) plays a vital role in regulation of a wide range of genes in the hypothalamus, and aids in both activation and repression of transcription. Mutation of the gene leads to a severe neurodevelopmental disorder associated with hypotonia, spasticity, severe learning disability, delayed psychomotor development, and recurrent pulmonary infections in males. Impairment in the expression of MeCP2 protein results in various neurological disorders such as Rett syndrome and Autism.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R E Amir et al.
Nature genetics, 23(2), 185-188 (1999-10-03)
Rett syndrome (RTT, MIM 312750) is a progressive neurodevelopmental disorder and one of the most common causes of mental retardation in females, with an incidence of 1 in 10,000-15,000 (ref. 2). Patients with classic RTT appear to develop normally until
J D Lewis et al.
Cell, 69(6), 905-914 (1992-06-12)
Methylation of mammalian DNA can lead to repression of transcription and alteration of chromatin structure. Recent evidence suggests that both effects are the result of an interaction between the methylated sites and methyl-CpG-binding proteins (MeCPs). MeCP1 has previously been detected
MeCP2 is a transcriptional repressor with abundant binding sites in genomic chromatin.
Nan X
Cell, 88, 471-481 (1997)
Rett Syndrome Mutant Neural Cells Lacks MeCP2 Immunoreactive Bands.
Bueno C
PLoS ONE, 11 (2016)
Yu-Chiao Huang et al.
Plant molecular biology, 90(1-2), 127-135 (2015-11-02)
Sucrose synthase (SuS), which catalyzes the reversible conversion of sucrose and uridine diphosphate (UDP) into fructose and UDP-glucose, is a key enzyme in sucrose metabolism in higher plants. SuS belongs to family 4 of the glycosyltransferases (GT4) and contains an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service