Skip to Content
Merck
All Photos(1)

Key Documents

M9317

Sigma-Aldrich

Anti-MeCP2 antibody produced in rabbit

~0.6 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ARR
WB
clone:
polyclonal
species reactivity:
human
citations:
28
technique(s):
microarray: suitable
western blot: 0.5-1 μg/mL using nuclear extracts of the MCF7 breast carcinoma cell line.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

human

concentration

~0.6 mg/mL

technique(s)

microarray: suitable
western blot: 0.5-1 μg/mL using nuclear extracts of the MCF7 breast carcinoma cell line.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MECP2(4204)
mouse ... Mecp2(17257)
rat ... Mecp2(29386)

General description

Methyl-CpG binding protein 2 (MeCP2) possesses a methyl-CpG-binding domain (MBD) and a transcriptional repression domain.

Immunogen

synthetic peptide corresponding to the C-terminus of human MeCP2 (amino acids 465-478) conjugated to KLH. The sequence is conserved in rat and mouse.

Biochem/physiol Actions

MeCP2 can bind to a single symmetrically methylated CpG pair. It is also capable of binding to chromosomes at sites known to have methylated DNA. MeCP2 silences transcription by recruiting the histone deacetylase (HDAC) repressive machinery via recruitment of the Sin 3A corepressor thus removing acetyl groups from histones and consequently silencing genes.
Methyl-CpG binding protein 2 (MeCP2) has been shown be crucial for brain development. It is part of the methyl-CpG-binding proteins, which are involved in repression of gene expression by binding to methylated DNA. Mutations in the gene encoding MeCP2 have been associated with Rett syndrome (RTT).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph R Kurian et al.
Epigenetics, 2(3), 173-178 (2007-10-30)
Pervasive developmental disorder is a classification covering five related conditions including the neurodevelopmental disorder Rett syndrome (RTT) and autism. Of these five conditions, only RTT has a known genetic cause with mutations in Methyl-CpG-binding protein 2 (MeCP2), a global repressor
Gilda Stefanelli et al.
Scientific reports, 6, 28295-28295 (2016-06-22)
MeCP2 is a transcriptional regulator whose functional alterations are responsible for several autism spectrum and mental disorders. Post-translational modifications (PTMs), and particularly differential phosphorylation, modulate MeCP2 function in response to diverse stimuli. Understanding the detailed role of MeCP2 phosphorylation is
David P Stuss et al.
Nucleic acids research, 41(9), 4888-4900 (2013-04-06)
MeCP2 is a methyl-CpG-binding protein that is a main component of brain chromatin in vertebrates. In vitro studies have determined that in addition to its specific methyl-CpG-binding domain (MBD) MeCP2 also has several chromatin association domains. However, the specific interactions
Tugba G Kucukkal et al.
Biochemistry, 54(41), 6357-6368 (2015-09-30)
Rett syndrome causing missense mutations in the methyl-CpG-binding domain (MBD) of methyl CpG-binding protein 2 (MeCP2) were investigated both in silico and in vitro to reveal their effect on protein stability. It is demonstrated that the vast majority of frequently
Paolo Petazzi et al.
RNA biology, 10(7), 1197-1203 (2013-04-25)
Mecp2 is a transcriptional repressor protein that is mutated in Rett syndrome, a neurodevelopmental disorder that is the second most common cause of mental retardation in women. It has been shown that the loss of the Mecp2 protein in Rett

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service