Skip to Content
Merck
All Photos(5)

Key Documents

A9771

Sigma-Aldrich

Albumin–fluorescein isothiocyanate conjugate

protein bovine

Synonym(s):

FITC-albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010201
NACRES:
NA.32

biological source

protein from bovine

form

powder

extent of labeling

≥7 mol FITCmol albumin

technique(s)

ELISA: suitable

solubility

10 mM Tris pH 7.0: 10 mg/mL

application(s)

detection

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Albumin makes up 60% of the plasma protein, which is produced by the liver. It regulates colloid osmotic pressure. Albumin is implicated in the transportation, distribution and metabolism of fatty acids, thyroid hormones and drugs. It reduces weight gain, blocks pulmonary edema, decreases ascites and lowers tissue edema.

Application

Albumin−fluorescein isothiocyanate conjugate has been used:
  • as a control to compare the intracellular delivery efficiency of proteins with or without carrier
  • for the fluorescent visualization of proteins and cells
  • as a blocking agent in ELISA
  • as a chemical for brain perfusion and fixation
  • for in vivo capillary–epithelial permeability experiment
  • for confocal microscopy of right ventricular (RV) vasculature in rats

Preparation Note

Prepared from crystallized and lyophilized albumin (A 4378).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philbert S Tsai et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(46), 14553-14570 (2009-11-20)
It is well known that the density of neurons varies within the adult brain. In neocortex, this includes variations in neuronal density between different lamina as well as between different regions. Yet the concomitant variation of the microvessels is largely
George A Tanner et al.
American journal of physiology. Renal physiology, 296(6), F1269-F1278 (2009-04-03)
Many aspects of the glomerular filtration of macromolecules remain controversial, including the location of the major filtration barrier, the effects of electrical charge, and the reason the filtration barrier does not clog. We examined these issues in anesthetized Necturus maculosus
Georgette L Suidan et al.
PloS one, 3(8), e3037-e3037 (2008-08-30)
Disruption of the blood brain barrier (BBB) is a hallmark feature of immune-mediated neurological disorders as diverse as viral hemorrhagic fevers, cerebral malaria and acute hemorrhagic leukoencephalitis. Although current models hypothesize that immune cells promote vascular permeability in human disease
Alcohol and the Liver (2013)
Airway epithelial KIF3A regulates Th2 responses to aeroallergens
Giridhar PV, et al.
Journal of Immunology, 23(7), 1600926-1600926 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service