Skip to Content
Merck
All Photos(1)

Key Documents

H6041

Sigma-Aldrich

Interferon α 2A human

HumanKine®, ≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyopholized powder, suitable for cell culture

Synonym(s):

α 2A Interferon, Human Interferon α 2A, IFN-α 2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Interferon α 2A human, IFN-α 2A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.4 ng/mL EC50

quality

endotoxin tested

mol wt

monomer 16 kDa (glycosylated)

packaging

glass bottle of 10 μg (in glass bottle)

manufacturer/tradename

HumanZyme

technique(s)

cell culture | mammalian: suitable

impurities

≤1EU/mg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IFNA2(3440)

Looking for similar products? Visit Product Comparison Guide

General description

The IFNA2 (interferon alpha 2) gene is mapped to human chromosome 9p21.3.[1] It is considered to be a prototypic type I IFN subtype. The encoded protein contains 165 amino acid. Ifna2 protein structure possesses five α-helices, linked by two disulfide bonds.[2]

Application

Interferon α 2A human has been used to check its antiviral action against chikungunya virus.[3] It has also been used as a control to check the antiviral cytokine induction of Korean red ginseng.[4]

Biochem/physiol Actions

IFNA2 is a type 1 interferon which acts as the first line of defense, and offers protection against pathogenic invasion.[5]
Interferon α 2A (IFN-α 2A) belongs to the type I interferon protein family and is produced by many cell types in response to viral infection. It has antiproliferative and antitumor activities, as well as induction of apoptosis and the inflammatory response. IFN-α 2A enhances the expression of MHC proteins by increasing the presentation of antigen peptides and activates cytotoxic T cells.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine IFN-α 2A, expressed in human HEK 293 cells, is a glycosylated monomer with an apparent molecular mass of 16 kDa. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The activity is determined by the dose-dependent cytotoxicity of the human TF-1 cell line (human erythroleukemic indicator cell line)) ≤ 0.4.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of tumor suppressive microRNA-31 enhances TRADD/NF-kappaB signaling in glioblastoma
Rajbhandari R, et al.
Testing, 6(19), 17805-17805 (2015)
Evaluation of broad-spectrum antiviral compounds against chikungunya infection using a phenotypic screening strategy
Bonotto RM, et al.
F1000Research, 7 (2018)
IFNA2: The prototypic human alpha interferon
Paul F, et al.
Gene, 567(2), 132-137 (2015)
Gillian M Keating
Drugs, 69(18), 2633-2660 (2009-12-01)
Peginterferon-alpha-2a (40 kD) [Pegasys] comprises an inert, branched, 40 kD polyethylene glycol (PEG) moiety attached to interferon-alpha-2a. Subcutaneous peginterferon-alpha-2a (40 kD) is indicated for the treatment of adults with hepatitis B e antigen (HBeAg)-positive or -negative chronic hepatitis B who
Evaluation of broad-spectrum antiviral compounds against chikungunya infection using a phenotypic screening strategy
Bonotto RM, et al.
F1000Research, 7 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service