Skip to Content
Merck
All Photos(1)

Key Documents

SCP0135

Sigma-Aldrich

Endothelin β Receptor Agonist IRL 1620

Synonym(s):

Endothelin Agonist, Endothelin Receptor Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C86H116N17O27
Molecular Weight:
1819.94
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

solubility

1 mg/mL at in water (Add acetonitrile dropwise to solution in water, mixing until the peptide dissolves)

storage temp.

−20°C

Amino Acid Sequence

Suc-Asp-Glu-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp

Application

IRL-1620 is used as a specific endothelin-B receptor (ET(B)-R) agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gulati et al.
Arzneimittel-Forschung, 62(1), 14-17 (2012-02-15)
ETB receptor agonist, IRL-1620 (or SPI-1620) presently in US Phase 1 clinical trial, has been demonstrated to selectively and transiently increase tumor blood flow. The present study was conducted to determine the effect of IRL-1620 on radiation therapy in tumor
Manish S Lavhale et al.
Pharmacological research, 62(6), 489-499 (2010-09-10)
Clonidine decreases mean arterial pressure (MAP) by acting as an α(2)-adrenergic receptor (AR) agonist in the central nervous system; it also acts on peripheral α-ARs to produce vasoconstriction. Endothelin (ET) has been shown to modulate the action of ARs. The
Mary G Leonard et al.
Brain research, 1420, 48-58 (2011-10-01)
Endothelin and its receptors have long been considered therapeutic targets in the treatment of ischemic stroke. Recent studies indicate that ET(B) receptors may provide both vasodilatation and neuroprotection. The purpose of this study was to determine the effect of selectively
Fernando S Carneiro et al.
The journal of sexual medicine, 5(12), 2793-2807 (2008-10-01)
The cavernosal tissue is highly responsive to endothelin-1 (ET-1), and penile smooth muscle cells not only respond to but also synthesize ET-1. Considering that ET-1 is directly involved in end-organ damage in salt-sensitive forms of hypertension, we hypothesized that activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service