Skip to Content
Merck

D9063

KIMBLE Dounce tissue grinder set

7 mL complete

Synonym(s):

885300-0007, Dounce Tissue Grinder, Tissue Grinder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass

feature

autoclavable

manufacturer/tradename

Kimble® 885300-0007

pestle A clearance

0.0028-0.0047 in.

pestle B clearance

0.0008-0.0022 in.

working volume × L

7 mL × 125 mm

Features and Benefits

  • Manufactured from borosilicate glass 3.3_x000D_
  • Designed primarily for cellular work where the nucleus remains intact after homogenization._x000D_
  • All-glass construction_x000D_
  • Two pestles are supplied with each complete unit_x000D_
  • Large clearance pestle is used for the initial sample reduction_x000D_
  • Small clearance pestle is used to form the final homogenate_x000D_
  • Replacement components are available and completely interchangeable

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences


Still not finding the right product?

Explore all of our products under KIMBLE Dounce tissue grinder set




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library