Skip to Content
Merck
All Photos(1)

Key Documents

N5636

Sigma-Aldrich

3-Nitropropionic acid

≥97% (HPLC), powder or crystals, mitochondrial complex II inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2N(CH2)2CO2H
CAS Number:
Molecular Weight:
119.08
Beilstein:
1759889
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

3-Nitropropionic acid, ≥97%

biological source

synthetic

Quality Level

Assay

≥97%

form

powder or crystals

color

white to light yellow

mp

68-70 °C (lit.)

solubility

alcohol: may be light yellow 100 mg/mL

storage temp.

2-8°C

SMILES string

OC(CC[N+]([O-])=O)=O

InChI

1S/C3H5NO4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)

InChI key

WBLZUCOIBUDNBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3-Nitropropionic acid has been used:
  • to generate multiple system atropy-parkinsoniam (MSA-P) model in rat to evaluate its effect on embryonic striatal and mesencephalic grafts
  • to inject into the nigrostriatal dopaminergic pathway in rats in order to examine its specific effects on the dopamine system
  • as succinate dehydrogenase (SDH) inhibitor for LUHMES neurons and cell lines

Biochem/physiol Actions

Administering 3-nitropropionic acid (3-NP) to the inner ear can stimulate sensorineural hearing loss. It serves as an irreversible inhibitor of mitochondrial complex II.
Excitotoxin shown to cause brain lesions similar to those of Huntington′s disease.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouse Models of Dystonia
Animal Models of Movement Disorders (2005)
No functional effects of embryonic neuronal grafts on motor deficits in a 3-nitropropionic acid rat model of advanced striatonigral degeneration (multiple system atrophy)
Waldner R, et al.
Neuroscience, 102(3), 581-592 (2001)
Dual loss of succinate dehydrogenase (SDH) and complex I activity is necessary to recapitulate the metabolic phenotype of SDH mutant tumors
Lorendeau D, et al.
Metabolic engineering, 43, 187-197 (2017)
Specific lesions in the extrapyramidal system of the rat brain induced by 3-nitropropionic acid (3-NPA)
Pei G and Ebendal T
Experimental Neurology, 132(1), 105-115 (1995)
The protective effect of Egb 761 against 3-nitropropionic acid-induced hearing loss: the role of sirtuin 1
Chang M Y, et al.
Clinical and Experimental Otorhinolaryngology, 11(1), 9-9 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service