Skip to Content
Merck
All Photos(2)

Key Documents

D8938

Sigma-Aldrich

KIMBLE Dounce tissue grinder set

2 mL complete

Synonym(s):

885300-0002, Dounce, Dounce Homogenizer, Glass Tissue Grinder, KIMBLE Tissue Grinder, KONTES, Pestles, Tissue Grinder, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

glass

feature

autoclavable

manufacturer/tradename

Kimble® 885300-0002

pestle A clearance

0.0030-0.0050 in.

pestle B clearance

0.0005-0.0025 in.

working volume × L

2 mL × 60 mm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naomi Habib et al.
Nature neuroscience, 23(6), 701-706 (2020-04-29)
The role of non-neuronal cells in Alzheimer's disease progression has not been fully elucidated. Using single-nucleus RNA sequencing, we identified a population of disease-associated astrocytes in an Alzheimer's disease mouse model. These disease-associated astrocytes appeared at early disease stages and
Fang-Yuan Hu et al.
Frontiers in bioengineering and biotechnology, 8, 564057-564057 (2020-10-20)
Retina is a crucial tissue for capturing and processing light stimulus. It is critical to describe the characteristics of retina at the single-cell level for understanding its biological functions. A variety of abnormalities in terms of morphology and function are
Sripriya Ravindra Kumar et al.
Nature methods, 17(5), 541-550 (2020-04-22)
Recombinant adeno-associated viruses (rAAVs) are efficient gene delivery vectors via intravenous delivery; however, natural serotypes display a finite set of tropisms. To expand their utility, we evolved AAV capsids to efficiently transduce specific cell types in adult mouse brains. Building
Jonathan M Levy et al.
Nature biomedical engineering, 4(1), 97-110 (2020-01-16)
The success of base editors for the study and treatment of genetic diseases depends on the ability to deliver them in vivo to the relevant cell types. Delivery via adeno-associated viruses (AAVs) is limited by AAV packaging capacity, which precludes
Alan Selewa et al.
Scientific reports, 10(1), 1535-1535 (2020-02-01)
A comprehensive reference map of all cell types in the human body is necessary for improving our understanding of fundamental biological processes and in diagnosing and treating disease. High-throughput single-cell RNA sequencing techniques have emerged as powerful tools to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service