Skip to Content
Merck
All Photos(3)

Documents

SAB3500181

Sigma-Aldrich

Anti-ADAM10 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KUZ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 85 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM10(102)

General description

ADAM metallopeptidase domain 10 or A disintegrin and metalloprotease 10 (ADAM10) is a transmembrane protein which is widely distributed. It consists of an amino-terminal signal sequence, a prodomain, a metalloprotease domain, a disintegrin domain, a cysteine-rich region, a transmembrane region and a cytoplasmic tail. The gene encoding this protein is localized on human chromosome 15q21.3.

Immunogen

ADAM10 antibody was raised against a peptide corresponding to amino acids 732 to 748 of human ADAM10. This sequence is identical to those of bovine and rat origins and differs from that of mouse ADAM10 by one amino acid (2,4).

Biochem/physiol Actions

ADAM metallopeptidase domain 10 (ADAM10) activates Notch proteins and has a role in embryonic development. It functions as a molecular scissor and cleaves the extracellular domains of proteins. The protein is a therapeutic target for a variety of diseases and malignancies.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500181.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duplication of the ALDH1A2 gene in association with pentalogy of Cantrell: a case report
Matthew B
Journal of Medical Case Reports, 7, 287-287 (2013)
Jialong Guo et al.
International journal of molecular medicine, 42(1), 643-650 (2018-04-26)
miR‑152 has been reported to be downregulated in rheumatoid arthritis (RA). However, the functional significance and molecular mechanisms underlying the role of miR‑152 in RA remain largely unknown. The present study aimed to explore the functional role and the underlying
Scissor sisters: regulation of ADAM10 by the TspanC8 tetraspanins.
Matthews AL
Biochemical Society Transactions, 45(3), 719-730 (2017)
Xin Li et al.
American journal of translational research, 10(12), 4054-4064 (2019-01-22)
Background: Pericardial patches are frequently used in vascular surgery to close arteriotomies. The early healing of these patches is mediated by attraction of CD34 and ephrinB2-positive cells. Atorvastatin is a commonly used statin drug that promotes healing of cardiovascular injury.
Corina Beerli et al.
Nature microbiology, 4(2), 216-225 (2018-11-14)
Cell motility is essential for viral dissemination1. Vaccinia virus (VACV), a close relative of smallpox virus, is thought to exploit cell motility as a means to enhance the spread of infection1. A single viral protein, F11L, contributes to this by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service