Skip to Content
Merck
All Photos(7)

Key Documents

A5316

Sigma-Aldrich

Anti-β-Actin (ACTB) Antibody

mouse monoclonal, AC-74

Synonym(s):

Actin Antibody Sigma, Beta Actin Antibody, Beta Actin Antibody - Monoclonal Anti-β-Actin antibody produced in mouse, Beta Actin Antibody Sigma, Sigma Beta Actin Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AC-74, monoclonal

mol wt

antigen 42 kDa

contains

15 mM sodium azide

species reactivity

rabbit, sheep, cat, guinea pig, bovine, mouse, chicken, wide range, Hirudo medicinalis, canine, Drosophila, carp, pig, rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: 1:1,000 using cultured human or chicken fibroblasts
western blot: 1:5,000 using cultured human or chicken fibroblast cell extracts

isotype

IgG2a

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)
mouse ... Actb(11461)
rat ... Actb(81822)

Looking for similar products? Visit Product Comparison Guide

General description

Actin is a highly conserved protein that is a major component of both the cytoskeletal and contractile structures in all cell types. It varies in amount, being related to the type of differentiation and to the functional state of cells and tissues. Actin can be found in two different forms of aggregation, the globular or the fibrillar state, and at least six distinct isoforms occur in vertebrates. The actins exhibit over 90% sequence homology, but each isoform has a unique NH2-terminal sequence. The isoforms are comprised of three α actins (skeletal, cardiac, smooth), one β actin (β-non-muscle) and two γ actins (γ smooth muscle and γ non-muscle). ACTB (actin β) gene codes for β-actin. It is a cytoskeletal housekeeping protein. It is present in the cytoplasm. The ACTB gene is located on human chromosome 7p22.1.
Monoclonal Anti-β-Actin (mouse IgG2a isotype) is derived from the AC-74 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Recognizes an epitope located on the N-terminal end of the β-isoform of actin. It specifically labels β-actin in a wide variety of tissues and species. In immunofluorescent staining of chicken gizzard ultrathin cryosections, the antibody labels the dense bodies, the longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. The antibody does not react with adult cardiac or skeletal muscle (besides traces due to contamination of the sample with non-muscle cells) or with β-actin expressing cells in Dictyostelium discoideum amoeba. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. B5, methacarn, ethanol or Bouin′s solutions may be also used as fixatives. The antibody can be used as probes for β-actin as an internal control in immunoblotting.

Immunogen

slightly modified β-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.

Application

Monoclonal mouse anti-actin antibody was used as a loading control for western blot analysis of immunoprecipitated proteins from rat dorsal root ganglion cocultures.
Monoclonal mouse anti-actin antibody was used as a loading control for western blot analysis of immunoprecipitated proteins from rat dorsal root ganglion cocultures.
Mouse Monoclonal Anti-β-Actin antibody has been used for western blot analyses. The antibody can also be used for immunohistochemistry, indirect immunofluorescence (1:1,000) using cultured human or chicken fibroblasts, and indirect ELISA.
Western blot analysis of MDCK cell lysates were performed using monoclonal anti-actin antibody as a primary antibody.
Monoclonal Anti-β-Actin antibody has been used in western blotting.
Mouse Monoclonal Anti-β-Actin antibody has been used for western blot analyses. The antibody can also be used for immunohistochemistry, indirect immunofluorescence (1:1,000) using cultured human or chicken fibroblasts, and indirect ELISA.
Western blot analysis of MDCK cell lysates were performed using monoclonal anti-actin antibody as a primary antibody.

Biochem/physiol Actions

The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are constituents of many cell types and are involved in a myriad of cellular processes including locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis. β-Actin (ACTB) modulates cell growth, migration and the G-actin pool. Mutations in ACTB leads to pleiotropic developmental disorder. It recognizes an epitope located on the N-terminal end of the β-isoform of actin. It specifically labels β-actin in a wide variety of tissues and species. In immunofluorescent staining of chicken gizzard ultrathin cryosections, the antibody labels the dense bodies, the longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. The antibody does not react with adult cardiac or skeletal muscle (besides traces due to contamination of the sample with non-muscle cells) or with β-actin expressing cells in Dictyostelium discoideum amoeba. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. B5, methacarn, ethanol or Bouin′s solutions may be also used as fixatives. The antibody can be used as probes for β-actin as an internal control in immunoblotting.

Physical form

The product is provided as ascites fluid containing 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional thermogenic beige adipogenesis is inducible in human neck fat.
Lee P, et al.
International Journal of Obesity, 38(2), 170-170 (2014)
Actin isoform compartments in chicken gizzard smooth muscle cells.
North A J, et al.
Journal of Cell Science, 107, 445-455 (1994)
Heparin-binding epidermal growth factor-like growth factor links hepatocyte priming with cell cycle progression during liver regeneration.
Mitchell C, et al.
The Journal of Biological Chemistry, 280(4), 2562-2568 (2005)
Myc confers androgen-independent prostate cancer cell growth.
Bernard D, et al.
The Journal of Clinical Investigation, 112(11), 1724-1731 (2003)
Lulu Huang et al.
Nucleic acid therapeutics, 29(4), 175-186 (2019-05-10)
Efforts to develop treatments for diseases caused by nonsense mutations have focused on identification of small molecules that promote translational read-through of messenger RNAs (mRNAs) harboring nonsense stop codons to produce full-length proteins. However, to date, no small molecule read-through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service