Skip to Content
Merck
All Photos(1)

Key Documents

AP132F

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, FITC conjugate

2 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Reacts with Rabbit IgG gamma chain, as well as the light chains from all Rabbit immunoglobulin classes. Monospecific by IEP when tested against whole Rabbit serum.

FLUOROPHORE/PROTEIN:

Approximately 12 mg/mg.

Application

Goat anti-Rabbit IgG Antibody, FITC conjugate is an antibody against Rabbit IgG for use in IF.

Physical form

Reconstitute in 1 mL of sterile distilled water.

Storage and Stability

that the concentration of protein and buffer salts will decrease to one-half of the original after

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danit Mechlovich et al.
The Journal of pharmacology and experimental therapeutics, 333(3), 874-882 (2010-03-20)
Increasing evidence suggests that oxidative stress (OS)-induced pancreatic beta-cell impairments is involved in diabetes and diabetic complications. Our group has recently synthesized two multifunctional nontoxic, lipophilic, iron-chelating drugs, 5-{N-methyl-N-propargylaminomethyl}-8-hydroxyquinoline (M30) and 5-{4-propargylpiperazin-1-ylmethyl}-8-hydroxyquinoline (HLA20), for the treatment of various OS-mediated pathogeneses.
Sean McCutcheon et al.
Cell adhesion & migration, 11(1), 1-12 (2016-01-09)
Central nervous system (CNS) cells cultured in vitro as neuroclusters are useful models of tissue regeneration and disease progression. However, the role of cluster formation and collective migration of these neuroclusters to external stimuli has been largely unstudied in vitro.
Qunhua Bai et al.
Experimental and therapeutic medicine, 12(2), 1060-1066 (2016-07-23)
Parkinson's disease (PD) is the second most common neurodegenerative disorder, which is characterized by the hallmark feature of loss of dopaminergic neurons in the substantia nigra. Energy metabolic disorder is associated with the pathogenesis of PD; however, the development of
Takahiko Akematsu et al.
Autophagy, 10(2), 209-225 (2013-11-28)
Programmed nuclear death (PND) in the ciliate protozoan Tetrahymena thermophila is a novel type of autophagy that occurs during conjugation, in which only the parental somatic macronucleus is destined to die and is then eliminated from the progeny cytoplasm. Other
STE20/SPS1-related proline/alanine-rich kinase is involved in plasticity of GABA signaling function in a mouse model of acquired epilepsy.
Yang, L; Cai, X; Zhou, J; Chen, S; Chen, Y; Chen, Z; Wang, Q; Fang, Z; Zhou, L
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service