Skip to Content
Merck
All Photos(2)

Documents

AB19026

Sigma-Aldrich

Anti-ADAM 10 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

A disintegrin and metalloproteinase domain 10, ADAM metallopeptidase domain 10, CD156c antigen, Kuzbanian protein homolog, Mammalian disintegrin-metalloprotease, a disintegrin and metalloprotease domain 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, mouse, human

species reactivity (predicted by homology)

rat

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

ADAM10 is a member of the ADAM (a disintegrin and metalloprotease like domain) family. It cleaves the membrane-bound precursor of TNF-alpha at 76-Ala-|-Val-77 to its mature soluble form. It is responsible for the proteolytic release of several cell-surface proteins, including heparin binding epidermal growth-like factor, ephrin-A2 and for constitutive and regulated alpha-secretase cleavage of amyloid precursor protein (APP). It contributes to the normal cleavage of the cellular prion protein and is involved in the cleavage of the adhesion molecule L1 at the cell surface and in released membrane vesicules, suggesting a vesicle-based protease activity.

Tumor-necrosis factor alpha is a proinflammatory cytokine and contributes to a variety of inflammatory disease responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNA-alpha, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10-, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloproteinase domain), was recently identified to cleave TNF-alpha, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM10, kuzbanian (KUZ), and MADM, respectively, (1,2,4). ADAM10mRNA is expressed in a variety of human and bovine tissues.

Specificity

Recognizes ADAM 10, C-terminus.

Immunogen

Epitope: a.a. 732-748 of human ADAM 10
Raised against a peptide corresponding to amino acids 732-748 of human ADAM 10 (Rosendahl, M.S., 1997). This sequence is identical to those of bovine and rat origins and differs from that of mouse ADAM 10 by one amino acid (Pan, D., 1997; Howard, L., 1996).

Application

Western blot:
1:500 - 1:2,000 dilution. Jurkat whole cell lysate can be used as a positive control and an 85 kDa band can be detected which may represent precursor. A 60 kDa faint band was detected in some cell lines including Jurkat, which appears to be the processed mature protein. Overloading of the primary antibody can lead to multiple bands appearing in some preparations. The presence of glycosylated forms and breakdown products can lead to other bands. A prominent proform at near 80-85 is usually seen, with bands at 60, and below 50 kDa in most cases. The immunogen peptide is available (Cat. No. AG533) for performing antibody blocking studies.

Optimal working dilutions must be determined by end user.

Immunohistochemistry:
A previous lot of this antibody was used to detect ADAM10 in formalin-fixed, paraffin-embedded renal tissue and renal cell carcinoma tissue (Gutwein, P., et al. EUROPEAN JOURNAL OF CANCER. 45(2009):478–489).

Immunofluorescence:
A previous lot of this antibody was used to detect ADAM10 in RCC4 (renal cell carcinoma) and A498 cells (Gutwein, P., et al. EUROPEAN JOURNAL OF CANCER. 45(2009):478–489).
This Anti-ADAM 10 Antibody, C-terminus is validated for use in WB for the detection of ADAM 10.

Quality

Routinely evaluated by Western Blot on RAW264.7 lysates.

Western Blot: 1:500 dilution of this lot detected ADAM 10 on 10 μg of RAW264.7 lysates

Target description

80-85 kDa

Linkage

Replaces: AB19031

Physical form

Format: Purified

Storage and Stability

Stable for 1 year at -20ºC in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
ADAM 10 is ubiquitously expressed. MDA-MB231 (Human breast cancer cell line known to express high levels of ADAM 10).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of E-cadherin in the pathogenesis of gastroesophageal reflux disease.
Jovov, B; Que, J; Tobey, NA; Djukic, Z; Hogan, BL; Orlando, RC
The American Journal of Gastroenterology null
Evidence for cadherin-11 cleavage in the synovium and partial characterization of its mechanism.
Noss, EH; Watts, GF; Zocco, D; Keller, TL; Whitman, M; Blobel, CP; Lee, DM; Brenner, MB
Arthritis Research & Therapy null
Anna Maria Kanarek et al.
The FEBS journal, 284(5), 742-753 (2017-01-20)
The association between hypertension and an increased risk for Alzheimer's disease (AD) and dementia is well established. Many data suggest that modulation of the renin-angiotensin system may be meaningful for the prevention and therapy of neurodegenerative disorders, in particular AD.
Xin Yan et al.
PloS one, 9(1), e84617-e84617 (2014-01-10)
Members of the ADAM (a disintegrin and metalloprotease) family are involved in embryogenesis and tissue formation via their proteolytic function, cell-cell and cell-matrix interactions. ADAM10 is expressed temporally and spatially in the developing chicken spinal cord, but its function remains
Xystus H L Tam et al.
Clinical science (London, England : 1979), 120(2), 81-89 (2010-08-24)
The sRAGE [soluble RAGE (receptor for advanced glycation end-products)] lack the transmembrane and cytoplasmic domain of the full-length receptor and can function as a decoy for RAGE ligands. Recent evidence suggests that sRAGE may be a potential biomarker of RAGE-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service